Skip to Content
Merck
All Photos(1)

Key Documents

246530

Sigma-Aldrich

Ethyl trifluoromethanesulfonate

99%

Synonym(s):

Ethyl 1,1,1-trifluoromethanesulfonate, Ethyl triflate, Ethyl trifluoromethanesulphonate, Trifluoromethanesulfonic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3SO2OC2H5
CAS Number:
Molecular Weight:
178.13
Beilstein:
1770746
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

99%

form

liquid

refractive index

n20/D 1.336 (lit.)

bp

115 °C (lit.)

density

1.374 g/mL at 25 °C (lit.)

functional group

fluoro
triflate

SMILES string

CCOS(=O)(=O)C(F)(F)F

InChI

1S/C3H5F3O3S/c1-2-9-10(7,8)3(4,5)6/h2H2,1H3

InChI key

UVECLJDRPFNRRQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl trifluoromethanesulfonate (EtOTf) is a useful reagent used in organic synthesis that can be used to introduce an ethyl group into a variety of compounds. EtOTf is also a good leaving group, which makes it useful in a variety of other reactions, such as nucleophilic substitutions and eliminations.

Application

Ethyl trifluoromethanesulfonate can be used as a precursor to prepare ionic liquids with triflate anions. It is reacted with a selected amine in the presence of a base, resulting in the formation of the desired ionic liquid with a triflate anion.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

95.0 °F - closed cup

Flash Point(C)

35 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service