Skip to Content
Merck
All Photos(1)

Key Documents

245720

Sigma-Aldrich

Diethyl adipate

ReagentPlus®, 99%

Synonym(s):

Hexanedioic acid 1,6-diethyl ester, NSC 19160, NSC 3363

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OCO(CH2)4COOC2H5
CAS Number:
Molecular Weight:
202.25
Beilstein:
1780035
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

product line

ReagentPlus®

Assay

99%

form

liquid

refractive index

n20/D 1.427 (lit.)

bp

251 °C (lit.)

mp

−20-−19 °C (lit.)

density

1.009 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)CCCCC(=O)OCC

InChI

1S/C10H18O4/c1-3-13-9(11)7-5-6-8-10(12)14-4-2/h3-8H2,1-2H3

InChI key

VIZORQUEIQEFRT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diethyl adipate (DEA), a fatty ester, is processed from the esterification of adipic acid and ethanol. It can be used as a plasticizer.

Application

DEA can be used in the synthesis of oligoadipamide, which can be layered with perfluoropolyether networks for protective coating applications. It can also be used as a plasticizer in the development of ethylene chlorotrifluoroethylene copolymer (ECTFE) based membranes for the pervaporation of water and toluene based blends.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pickering emulsions for skin decontamination
Salerno A, et al.
Toxicology in vitro, 34, 45-54 (2016)
Oligoamide grafted with perfluoropolyether blocks: A potential protective coating for stone materials
Cao Y, et al.
Progress in Organic Coatings, 111(8), 164-174 (2017)
Preparation and characterization of ECTFE solvent resistant membranes and their application in pervaporation of toluene/water mixtures
Simone S, et al.
Separation and Purification Technology, 90(8), 147-161 (2012)
Differential determination of plasticizers and organophosphorus flame retardants in residential indoor air in Japan
Takeuchi S, et al.
Environmental Science and Pollution Research International, 25(8), 7113-7120 (2018)
Characterization of microporous ECTFE membrane after exposure to different liquid media and radiation
Yao N, et al.
Journal of Membrane Science , 532(8), 89-104 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service