Skip to Content
Merck
All Photos(1)

Key Documents

241288

Sigma-Aldrich

Triphenyl phosphate

≥99%

Synonym(s):

Triphenoxyphosphine oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5O)3PO
CAS Number:
Molecular Weight:
326.28
Beilstein:
1888236
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

1.3 mmHg ( 200 °C)

Quality Level

Assay

≥99%

bp

244 °C/10 mmHg (lit.)

mp

48-50 °C (lit.)

solubility

acetone: soluble(lit.)
alcohol: moderately soluble(lit.)
benzene: soluble(lit.)
chloroform: soluble(lit.)
diethyl ether: soluble(lit.)
water: insoluble(lit.)

functional group

phosphate

SMILES string

O=P(Oc1ccccc1)(Oc2ccccc2)Oc3ccccc3

InChI

1S/C18H15O4P/c19-23(20-16-10-4-1-5-11-16,21-17-12-6-2-7-13-17)22-18-14-8-3-9-15-18/h1-15H

InChI key

XZZNDPSIHUTMOC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Triphenyl phosphate, an organophosphorus flame retardant (PFR), was determined in human milk sample.

Application

Triphenyl phosphate was used as an internal standard in the screening and quantification of pesticide residues in vegetables and fruits.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

428.0 °F - closed cup

Flash Point(C)

220 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joon-Woo Kim et al.
Chemosphere, 116, 91-97 (2014-03-19)
In this study, the concentrations of 10 organophosphorus flame retardants (PFRs) were determined in 89 human breast milk samples collected from Japan, the Philippines and Vietnam. Among the targeted PFRs, tris(2-chloroexyl) phosphate (TCEP) and triphenyl phosphate (TPHP) were the predominant
Felix D Guerrero et al.
Journal of medical entomology, 43(5), 896-901 (2006-10-05)
The horn fly, Haematobia irritans irritans (L.) (Diptera: Muscidae), has become a problem for Brazilian cattle producers even though its introduction into Brazil is relatively recent. Failure to control this cattle pest is becoming a concern, and horn fly populations
M I Picollo et al.
Journal of medical entomology, 37(5), 721-725 (2000-09-27)
Permethrin-resistant colonies of Pediculus capitis (De Geer) from Buenos Aires were used to establish a resistance profile and to examine resistance mechanisms. All permethrin-resistant head lice (resistance ratio from 52.8 to > 88.7) were also resistant to d-phenothrin (resistance ratio
Rixin Lu et al.
Journal of hazardous materials, 243, 311-318 (2012-11-07)
Liquid crystal display (LCD) glass mainly consists of polarizing film, liquid crystal and glass substrate. Removing and recovering the liquid crystal and films from the LCD glass effectively has important significance for recovering the other parts. This study proposed a
Amer El-Batta et al.
The Journal of organic chemistry, 72(14), 5244-5259 (2007-06-15)
Water is demonstrated to be an effective medium for the Wittig reaction over a wide range of stabilized ylides and aldehydes. Despite sometimes poor solubility of the reactants, good chemical yields normally ranging from 80 to 98% and high E-selectivities

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service