Skip to Content
Merck
All Photos(1)

Key Documents

239356

Sigma-Aldrich

N,N-Dimethylethylamine

99%

Synonym(s):

N-Ethyldimethylamine, DMEA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NC2H5
CAS Number:
Molecular Weight:
73.14
Beilstein:
1696893
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

8.09 psi ( 20 °C)

Quality Level

Assay

99%

refractive index

n20/D 1.372 (lit.)

bp

36-38 °C (lit.)

mp

−140 °C (lit.)

density

0.675 g/mL at 25 °C (lit.)

SMILES string

CCN(C)C

InChI

1S/C4H11N/c1-4-5(2)3/h4H2,1-3H3

InChI key

DAZXVJBJRMWXJP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N,N-Dimethylethylamine was used in the preparation of pH and reduction dual-responsive polypeptide nanogels with self-reinforced endocytoses.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-18.4 °F - closed cup

Flash Point(C)

-28 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jianxun Ding et al.
Journal of controlled release : official journal of the Controlled Release Society, 172(2), 444-455 (2013-06-08)
The pH and reduction dual-responsive polypeptide nanogels with self-reinforced endocytoses were prepared through ring-opening polymerization of l-glutamate N-carboxyanhydrides, deprotection of benzyl group and subsequent quaternization reaction between γ-2-chloroethyl-l-glutamate unit in polypeptide block and 2,2'-dithiobis(N,N-dimethylethylamine). The nanogels were revealed to exhibit
T Lundh et al.
British journal of industrial medicine, 48(3), 203-207 (1991-03-01)
The exposure and metabolism of dimethylethylamine (DMEA) was studied in 12 mould core makers in four different foundries using the Ashland cold box technique. The mean time weighted average (TWA) full work shift DMEA exposure concentration was 3.7 mg/m3. Inhaled
J A Post et al.
The American journal of physiology, 268(2 Pt 2), H773-H780 (1995-02-01)
Phosphatidylethanolamine (PE) is a nonbilayer-preferring and fusogenic phospholipid. It is kept in the bilayer configuration by interaction with other phospholipids in biologic membranes. However, reorganization of the membrane phospholipids could lead to expression of the nonbilayer nature of PE and
L Hansén et al.
Scandinavian journal of work, environment & health, 11(4), 307-310 (1985-08-01)
A method for the determination of dimethylethylamine in workroom air has been developed. Static gas standards of the amine in air were used to evaluate different sampling techniques. The analysis was performed by isotachophoresis. The sampling equipment of choice was
T Lundh et al.
Occupational and environmental medicine, 52(7), 478-483 (1995-07-01)
The aim was to study the effect of trimethylamine (TMA) on the metabolism of the industrial catalyst dimethylethylamine (DMEA) to ascertain whether biological monitoring of industrial exposure to DMEA is compromised and excretion of the malodorous DMEA in sweat and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service