Skip to Content
Merck
All Photos(1)

Key Documents

232297

Sigma-Aldrich

Benzenesulfonyl isocyanate

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5SO2NCO
CAS Number:
Molecular Weight:
183.18
Beilstein:
743441
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
liquid
Assay:
95%

Quality Level

Assay

95%

form

liquid

refractive index

n20/D 1.536 (lit.)

bp

130 °C/9 mmHg (lit.)

density

1.369 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

O=C=NS(=O)(=O)c1ccccc1

InChI

1S/C7H5NO3S/c9-6-8-12(10,11)7-4-2-1-3-5-7/h1-5H

InChI key

UJYAZVSPFMJCLW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Benzenesulfonyl isocyanate reacts with ethanol and phenol to yield normal urethan products.

Application

Benzenesulfonyl isocyanate was used in synthesis of sterically hindered derivatives of phenols and alcohols. It was also used in the reaction of semi crystalline poly(ether ether ketone) films with carbonyl selective reagents to produce modified film samples.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

The Chemistry of Benzenesulfonyl Isocyanate. I. Identification of Hindered Phenols and Alcohols1.
McFarland JW and Howard JB.
The Journal of Organic Chemistry, 30(3), 957-958 (1965)
Surface modifications of poly (ether ether ketone).
Franchina NL and McCarthy TJ.
Macromolecules, 24(11), 3045-3049 (1991)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service