Skip to Content
Merck
All Photos(3)

Key Documents

228672

Sigma-Aldrich

Samarium(III) oxide

99.9% trace metals basis

Synonym(s):

Samaria

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Sm2O3
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Quality Level

Assay

99.9% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: samarium

density

8.35 g/mL at 25 °C (lit.)

SMILES string

O=[Sm]O[Sm]=O

InChI

1S/3O.2Sm

InChI key

PRCWVHVINXAFRG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lloyd G Tillman et al.
Journal of pharmaceutical sciences, 97(1), 225-236 (2007-08-28)
Treatment of systemic disease with phosphorothioate antisense oligonucleotides (PS ASOs) has been accomplished using local or parenteral routes of administration to date. This report describes, for the first time, the effective oral delivery of a second generation oligonucleotide where significant
S F Ahrabi et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 8(3), 193-201 (1999-06-24)
The effect of the neutron activation factors, i.e., admixture of samarium oxide (Sm2O3) and irradiation time, on the physico-chemical properties of the raw materials and the in vitro dissolution and disintegration of hydrophilic and lipophilic suppositories was investigated. It was
Guohui Zhang et al.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine, 67(1), 46-49 (2008-08-19)
Cross sections of the (147)Sm(n, alpha)(144)Nd reaction were measured at En=5.0 and 6.0MeV. A twin gridded ionization chamber was used as a charged particle detector and two large area (147)Sm(2)O(3) samples placed back to back were employed. Experiments were performed
Matthew D Burke et al.
Pharmaceutical research, 24(4), 695-704 (2007-03-21)
To develop a robust radiolabeling technique to enable evaluation of difficult to radiolabel gastric retentive formulations using gamma scintigraphy. The use of a successful radiolabel will allow accurate assessment of the gastric residence time of the formulations. The retention of
B Mandal et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 60(4), 933-939 (2004-03-24)
Microphone based photoacoustic (PA) spectrometer to study solids in powder form was designed and fabricated. Laser PA spectra of Sm3+ ion in Sm2O3 and SmCl3.6H2O microcrystalline powders were recorded first time in 484-542 nm spectral region at room temperature. Analysis

Questions

  1. What size particle is the Sm2O3

    1 answer
    1. Unfortunately, the particle size of this product is not determined.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service