Skip to Content
Merck
All Photos(1)

Key Documents

215147

Sigma-Aldrich

Calcium

granular, 99%

Synonym(s):

Calcium atom, Calcium element

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ca
CAS Number:
Molecular Weight:
40.08
Beilstein:
4241647
EC Number:
MDL number:
UNSPSC Code:
12141503
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

99%

form

granular

reaction suitability

reagent type: reductant

resistivity

3.5 μΩ-cm, 20°C

bp

1484 °C (lit.)

mp

850 °C (lit.)

density

1.54 g/mL at 25 °C (lit.)

SMILES string

[Ca]

InChI

1S/Ca

InChI key

OYPRJOBELJOOCE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Calcium is a highly reactive and non-toxic metal often prepared by the aluminothermal or electrolysis methods. Calcium metal granules with a purity of 99% are a valuable material used in several industrial and academic applications. The granules are made from high-quality calcium and are produced under strict manufacturing standards to ensure their purity and quality. The granular form allows for easier handling and storage than a powder, making it convenient for use in various applications.

Application

Our 99% pure calcium metal granules are widely used as a reducing agent in the manufacturing of various metals, including uranium and thorium. They are also an essential component in the production of aluminum alloys that are commonly used in the aerospace and automotive industries. These calcium metal granules are also used in the desulfurization of steel and as a deoxidizer in the production of various metals. Furthermore, our calcium metal granules are extensively used in organic synthesis to reduce various functional groups, such as aldehydes, ketones, esters, and nitriles, to their corresponding alcohols. The metal is also utilized in the preparation of Grignard reagents, which serve as crucial intermediates in organic synthesis.
Precursor to Ca8Al3 and Ca13Al14; intermetallics with new metallic Zintl phases.7

Features and Benefits

  • Quality assured - made from high-quality raw materials and produced under strict manufacturing standards to ensure consistent performance.
  • Easier to handle and store - in comparison to powder, the granular form ensures higher stability during storage and handling of materials.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Water-react 2

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eleni Gourni et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 55(10), 1719-1725 (2014-08-26)
Gastrin-releasing peptide receptors (GRPrs) are overexpressed on a variety of human cancers, providing the opportunity for peptide receptor targeting via radiolabeled bombesin-based peptides. As part of our ongoing investigations into the development of improved GRPr antagonists, this study aimed at
Xinyu Zhou et al.
Circulation research, 114(4), 706-716 (2014-02-15)
Trimeric intracellular cation channels (TRIC) represents a novel class of trimeric intracellular cation channels. Two TRIC isoforms have been identified in both the human and the mouse genomes: TRIC-A, a subtype predominantly expressed in the sarcoplasmic reticulum (SR) of muscle
Jun Suzuki et al.
Science (New York, N.Y.), 341(6144), 403-406 (2013-07-13)
A classic feature of apoptotic cells is the cell-surface exposure of phosphatidylserine (PtdSer) as an "eat me" signal for engulfment. We show that the Xk-family protein Xkr8 mediates PtdSer exposure in response to apoptotic stimuli. Mouse Xkr8(-/-) cells or human
Tsai-Wen Chen et al.
Nature, 499(7458), 295-300 (2013-07-23)
Fluorescent calcium sensors are widely used to image neural activity. Using structure-based mutagenesis and neuron-based screening, we developed a family of ultrasensitive protein calcium sensors (GCaMP6) that outperformed other sensors in cultured neurons and in zebrafish, flies and mice in
Byrappa Venkatesh et al.
Nature, 505(7482), 174-179 (2014-01-10)
The emergence of jawed vertebrates (gnathostomes) from jawless vertebrates was accompanied by major morphological and physiological innovations, such as hinged jaws, paired fins and immunoglobulin-based adaptive immunity. Gnathostomes subsequently diverged into two groups, the cartilaginous fishes and the bony vertebrates.

Articles

Magnetic nanoparticles have attracted tremendous attention due to their novel properties and their potential applications in magnetic recording, magnetic energy storage and biomedicine.

Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. Hi ! What is the size of the granules ? Thank you !

    1 answer
    1. Unfortunately, the granules size of this product is not determined.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service