Skip to Content
Merck
All Photos(2)

Key Documents

214736

Sigma-Aldrich

Xylenes

reagent grade

Synonym(s):

Xylene mixture of isomers

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(CH3)2
CAS Number:
Molecular Weight:
106.17
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.21
grade:
reagent grade
bp:
137-140 °C (lit.)
vapor pressure:
18 mmHg ( 37.7 °C)

grade

reagent grade

vapor density

3.7 (vs air)

vapor pressure

18 mmHg ( 37.7 °C)

Assay

≥75.0% (xylenes isomers)

form

liquid

autoignition temp.

867 °F

expl. lim.

7 %

dilution

(for analytical testing)

impurities

≤25% ethylbenzene

refractive index

n20/D 1.497 (lit.)

bp

137-140 °C (lit.)

mp

-34 °C (lit.)

density

0.86 g/mL at 25 °C (lit.)

SMILES string

Cc1ccc(C)cc1.Cc2cccc(C)c2.Cc3ccccc3C

InChI

1S/3C8H10/c1-7-3-5-8(2)6-4-7;1-7-4-3-5-8(2)6-7;1-7-5-3-4-6-8(7)2/h3*3-6H,1-2H3

InChI key

MVZVDAGWAAZJPE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Xylenes is the mixture of isomeric ortho-, meta- and para-forms of xylene. Its dispersed emission and time of flight mass spectra has been reported. Transitions from S1 ↔ S0 states have been found to be within the range of 100cm-1 and were reported to be arising due to the internal rotation of ring methyl groups. Time-resolvedin situ FTIR spectroscopic and gas chromatographic studies have been carried out to study its diffusion coefficients, adsorption rates and isomerization efficacy over HSZM-5 surfaces.

Application

Xylenes may be employed in chloramphenicol acetyltransferase (CAT) assay to evaluate the CAT gene expression.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system,Liver,Kidney, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

77.0 °F - closed cup

Flash Point(C)

25 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

A study of nonrigid aromatic molecules by supersonic molecular jet spectroscopy. I. Toluene and the xylenes.
Breen PJ, et al.
J. Chem. Phys. , 87(4), 1917-1926 (1987)
Transport and isomerization of xylenes over HZSM-5 zeolites.
Mirth G, et al.
J. Catal., 139(1), 24-33 (1993)
K J Li et al.
Proceedings of the National Academy of Sciences of the United States of America, 95(7), 3650-3654 (1998-05-09)
Efficient and controllable expression of a transgene usually requires the presence of intron sequences and much efforts have been made to produce retrovirus vectors that can transduce and integrate genes with introns. However, this has proven difficult because the viral
Chani M Becker et al.
Neuro-oncology, 17(9), 1210-1219 (2015-05-15)
Targeting drug delivery to invasive glioma cells is a particularly difficult challenge because these cells lie behind an intact blood-brain barrier (BBB) that can be observed using multimodality imaging. BBB-associated efflux transporters such as P-glycoprotein (P-gp) and breast cancer resistance
Anne-France le Rolle et al.
Journal of translational medicine, 13, 199-199 (2015-06-25)
Clinical therapy for metastatic colorectal cancer (CRC) remains limited, especially when the tumor harbors a KRAS mutation. This study aimed to identify prognostic biomarkers in CRC that are accessible for therapeutic inhibition. Conditioned media from human CRC epithelial cells and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service