Skip to Content
Merck
All Photos(1)

Key Documents

213969

Sigma-Aldrich

(−)-Diethyl D-tartrate

≥99%

Synonym(s):

D-(−)-Tartaric acid diethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CCCH(OH)CH(OH)CO2H
CAS Number:
Molecular Weight:
206.19
Beilstein:
1727143
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥99%

optical activity

[α]23/D −8.5°, neat

optical purity

ee: ≥99% (GLC)

refractive index

n20/D 1.446 (lit.)

bp

162 °C/19 mmHg (lit.)

density

1.205 g/mL at 20 °C (lit.)

functional group

ester
hydroxyl

SMILES string

CCOC(=O)[C@@H](O)[C@H](O)C(=O)OCC

InChI

1S/C8H14O6/c1-3-13-7(11)5(9)6(10)8(12)14-4-2/h5-6,9-10H,3-4H2,1-2H3/t5-,6-/m0/s1

InChI key

YSAVZVORKRDODB-WDSKDSINSA-N

Looking for similar products? Visit Product Comparison Guide

Application

(-)-Diethyl D-tartrate can be used as a starting material in the synthesis of bioactive compounds such as (-)-kumausallene, syringolide 2 and (+)-exo-brevicomin.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

200.1 °F - closed cup

Flash Point(C)

93.4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Synthesis and absolute configuration of syringolide 2, an elicitor from Pseudomonas syringae pv. tomato.
Kuwahara S, et al.
Tetrahedron Letters, 36(18), 3201-3202 (1995)
A new expeditious synthesis of (+)-exo-brevicomin via efficient C? C bond formation of triflates.
Kotsuki H, et al.
Tetrahedron Letters, 30(30), 3999-4000 (1989)
Radical cyclization of ?-alkoxyacrylates: A formal synthesis of (-)-kumausallene.
Lee E, et al.
Tetrahedron Letters, 39(3), 317-318 (1998)
Kimberly A Kahle et al.
Electrophoresis, 28(15), 2644-2657 (2007-06-29)
In this second study on dual-chirality microemulsions containing a chiral surfactant and a chiral oil, a less hydrophobic and lower interfacial tension chiral oil, diethyl tartrate, is employed (Part 1, Foley, J. P. et al.., Electrophoresis, DOI: 10.1002/elps.200600551). Six stereochemical
Total synthesis of the light-harvesting carotenoid peridinin.
Thomas Olpp et al.
Angewandte Chemie (International ed. in English), 45(24), 4023-4027 (2006-05-10)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service