213721
1,2-Dodecanediol
90%
Synonym(s):
1,2-Dihydroxydodecane, 1,2-Dodecylene glycol, 2-Hydroxylauryl alcohol, n-Dodecane-1,2-diol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3(CH2)9CH(OH)CH2OH
CAS Number:
Molecular Weight:
202.33
Beilstein:
1700179
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
90%
mp
56-60 °C (lit.)
functional group
hydroxyl
SMILES string
CCCCCCCCCCC(O)CO
InChI
1S/C12H26O2/c1-2-3-4-5-6-7-8-9-10-12(14)11-13/h12-14H,2-11H2,1H3
InChI key
ZITKDVFRMRXIJQ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
1,2-Dodecanediol was used in the preparation of:
- lipophilic diamines and amino alcohols
- Fe(II), Co(II) and Ni(II) ferrite nanoparticles in the range of 9-25 nm
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Fei Gong et al.
Small (Weinheim an der Bergstrasse, Germany), 16(46), e2003496-e2003496 (2020-10-28)
Nanozymes with high catalytic activity and great stability have attracted increasing interests as the promising alternative to natural enzymes for applications in various fields. In this study, a new type of highly efficient peroxidase-like nanozymes based on FeWOX nanosheets (NSs)
Bo Zhang et al.
Nanoscale, 9(47), 18711-18722 (2017-11-23)
Recently, immunochromatography test strips (ICTS) have been fully developed for point-of-care testing (POCT). However, the intrinsic limitations including non-quantitative detection of colloidal gold ICTS and low sensitivity of fluorescence ICTS (FICTS) significantly restrict their further application in clinical diagnosis. Taking
Xiaoqun Gong et al.
Nanomedicine : nanotechnology, biology, and medicine, 14(4), 1257-1266 (2018-04-01)
Immunochromatography test strip (ICTS) displayed high advantages in screening acute myocardial infarction (AMI) biomarkers. However, the low sensitivity and nonquantitative results seriously limited its clinical application. Herein, we designed a highly sensitive, quantitative and dual-readout ICTS for assaying multiple AMI
Celso O R Júnior et al.
Memorias do Instituto Oswaldo Cruz, 104(5), 703-705 (2009-10-13)
A series of diamines and amino alcohols derived from 1-dodecanol, 1-tetradecanol, 1,2-dodecanediol and 1,2-tetradecanediol were synthesized and tested for their antitubercular activity. Compounds 3, 8 and 9 were found to be the most active (MIC of 6.25 microg/mL). Nine other
Karine Priscila Naidek et al.
Journal of colloid and interface science, 358(1), 39-46 (2011-03-26)
We synthesized magnetic spinel ferrites from trimetallic single-source precursors. Fe(II), Co(II), and Ni(II) ferrite nanoparticles in the range of 9-25 nm were synthesized by solvothermal decomposition of trimetallic acetate complex precursors in benzyl ether in the presence of oleic acid
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service