Skip to Content
Merck
All Photos(2)

Key Documents

204501

Sigma-Aldrich

Ammonium sulfate

99.999% trace metals basis

Synonym(s):

Mascagnite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)2SO4
CAS Number:
Molecular Weight:
132.14
EC Number:
MDL number:
UNSPSC Code:
12352302
eCl@ss:
38030408
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
crystals and lumps
solubility:
acetone: insoluble(lit.)
ethanol: insoluble(lit.)
Pricing and availability is not currently available.

grade

for analytical purposes

Assay

99.999% trace metals basis

form

crystals and lumps

technique(s)

HPLC: suitable

impurities

≤15.0 ppm Trace Metal Analysis

pH

5.0-6 (25 °C, 132 g/L)

mp

>280 °C (dec.) (lit.)

solubility

acetone: insoluble(lit.)
ethanol: insoluble(lit.)

density

1.77 g/mL at 25 °C (lit.)

SMILES string

N.N.OS(O)(=O)=O

InChI

1S/2H3N.H2O4S/c;;1-5(2,3)4/h2*1H3;(H2,1,2,3,4)

InChI key

BFNBIHQBYMNNAN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Ammonium sulfate is a less corrosive sulfating agent that is used to synthesize the highly efficient acid catalyst for biodiesel production.

It can also be used to enhance the stability of perovskite solar cells. The incorporation of sulfate species minimizes chemical degradation at the perovskite/ETL interface. It is insoluble in acetone, alcohol and ether.

Application

Ammonium sulfate can be used:
  • As a structure-directing agent (SDA) to prepare hexagonal WO3 nanorod thin films for high-performance energy storage devices. The addition of (NH4)2SO4 prevents agglomeration of crystals.[1]
  • As an electrolyte for aqueous ammonium dual-ion batteries and MnO2/carbon supercapacitors.[2] [3]
  • As a blowing agent to prepare porous carbon structures for supercapacitors. These porous materials show high capacitance and long-term stability.[4]
  • To enhance the stability of perovskite solar cells. The incorporation of sulfate species minimizes chemical degradation at the perovskite/ETL interface.[5]

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A novel aqueous ammonium dual-ion battery based on organic polymers
Yadi Zhang, et al.
Journal of Materials Chemistry, 7, 11314-11320 (2019)
N, S co-doped porous graphene-like carbon synthesized by a facile coal tar pitch-blowing strategy for high-performance supercapacitors
Gang Li, et al.
Chemical Physics Letters, 827, 140712-140712 (2023)
Low temperature water based electrolytes for MnO 2/carbon supercapacitors
Alexander J Roberts, et al.
Physical Chemistry Chemical Physics, 15, 3518-3526 (2013)
Ammonium sulfate treatment at the TiO2/perovskite interface boosts operational stability of perovskite solar cells
Bening Tirta Muhammad, et al.
Journal of Materials Chemistry, 9, 14334-14341 (2021)
Facile synthesis of self-assembled WO3 nanorods for high-performance electrochemical capacitor
Pragati A. Shinde, et al.
Journal of alloys and compounds, 770, 1130-1137 (2019)

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service