Skip to Content
Merck
All Photos(3)

Key Documents

202983

Sigma-Aldrich

Cerium(III) chloride heptahydrate

99.999% trace metals basis

Synonym(s):

Cerous chloride heptahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CeCl3 · 7H2O
CAS Number:
Molecular Weight:
372.58
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.999% trace metals basis
form:
crystals and lumps

Quality Level

Assay

99.999% trace metals basis

form

crystals and lumps

reaction suitability

reagent type: catalyst
core: cerium

impurities

≤15.0 ppm Trace Metal Analysis

density

~3.94 g/mL at 25 °C (lit.)

SMILES string

[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].Cl[Ce](Cl)Cl

InChI

1S/Ce.3ClH.7H2O/h;3*1H;7*1H2/q+3;;;;;;;;;;/p-3

InChI key

KPZSTOVTJYRDIO-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Cerium chloride heptahydrate is often used to prepare cerium oxide nanoparticles by various methods. It reacts with sodium-oleate to form a cerium-oleate complex which can be further decomposed in hydrocarbon solvents to prepare cerium oxide nanoparticles. Cerium chloride heptahydrate can also be used in the conversion of esters to allylsilanes.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1C

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organic Syntheses, 69, 89-89 (1990)
Panthenol-stabilized cerium dioxide nanoparticles for cosmeceutic formulations against ROS-induced and UV-induced damage.
Zholobak NM, et al.
Journal of Photochemistry and Photobiology. B, Biology, 130, 102-108 (2014)
Preparation and characterization of monodisperse cerium oxide nanoparticles in hydrocarbon solvents
Gu H, et al.
Chemistry of Materials, 19.5, 1103-1110 (2007)
Cerium fluoride nanoparticles protect cells against oxidative stress.
Shcherbakov AB, et al.
Materials Science and Engineering, C, 50, 151-159 (2015)
Haiquan Zhao et al.
Archives of environmental contamination and toxicology, 62(1), 154-164 (2011-04-20)
Cerium has been demonstrated to damage liver of mice, but very little is known about the molecular mechanisms underlying the mouse liver apoptosis. In order to understand the liver injury induced by intragastric administration of cerium chloride (CeCl3) for 60

Articles

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service