Skip to Content
Merck
All Photos(4)

Key Documents

202738

Sigma-Aldrich

Barium chloride

99.999% trace metals basis

Synonym(s):

Barium dichloride, Barium(2+) dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BaCl2
CAS Number:
Molecular Weight:
208.23
EC Number:
MDL number:
UNSPSC Code:
12352302
eCl@ss:
38050210
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.999% trace metals basis
form:
powder and chunks

Quality Level

Assay

99.999% trace metals basis

form

powder and chunks

impurities

≤15.0 ppm Trace Metal Analysis

mp

963 °C (lit.)

density

3.856 g/mL at 25 °C (lit.)

SMILES string

Cl[Ba]Cl

InChI

1S/Ba.2ClH/h;2*1H/q+2;;/p-2

InChI key

WDIHJSXYQDMJHN-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Barium chloride (BaCl2) is a water soluble ionic compound that is hygroscopic in nature. It can be found naturally as witherite and can be prepared from barium hydroxide or barium carbonate.

Application

BaCl2 is an important compound that is used in a variety of industrial and laboratory based applications. Its other applications include testing of sulfate ions, pigmet synthesis and pharmaceuticals.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Eye Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Group 17 (H, F, Cl, Br, I) Alkaline Earth Compounds
Encyclopedia of the Alkaline Earth Compounds, 25-104 (2013)
Role of M 3 receptor in aconitine/barium-chloride-induced preconditioning against arrhythmias in rats
Liu Y, et al.
Naunyn-Schmiedeberg'S Archives of Pharmacology, 379(5), 511-515 (2009)
Alginate-based microencapsulation of retinal pigment epithelial cell line for cell therapy
Wikstrom J, et al.
Biomaterials, 29(7), 869-876 (2008)
Francesca Boscolo Sesillo et al.
Cell reports, 26(3), 689-701 (2019-01-17)
Most human cancers originate from high-turnover tissues, while low-proliferating tissues, like skeletal muscle, exhibit a lower incidence of tumor development. In Duchenne muscular dystrophy (DMD), which induces increased skeletal muscle regeneration, tumor incidence is increased. Rhabdomyosarcomas (RMSs), a rare and
David Sala et al.
Nature communications, 10(1), 1796-1796 (2019-04-19)
Metabolic reprogramming is an active regulator of stem cell fate choices, and successful stem cell differentiation in different compartments requires the induction of oxidative phosphorylation. However, the mechanisms that promote mitochondrial respiration during stem cell differentiation are poorly understood. Here

Articles

Three approaches generate white light, including LED-based down-conversion for broader applications.

Three approaches generate white light, including LED-based down-conversion for broader applications.

Three approaches generate white light, including LED-based down-conversion for broader applications.

Three approaches generate white light, including LED-based down-conversion for broader applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service