Skip to Content
Merck
All Photos(3)

Key Documents

201057

Sigma-Aldrich

Butyltin trichloride

95%

Synonym(s):

Butyltrichlorotin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3SnCl3
CAS Number:
Molecular Weight:
282.18
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

liquid

refractive index

n20/D 1.523 (lit.)

bp

93 °C/10 mmHg (lit.)

density

1.693 g/mL at 25 °C (lit.)

SMILES string

CCCC[Sn](Cl)(Cl)Cl

InChI

1S/C4H9.3ClH.Sn/c1-3-4-2;;;;/h1,3-4H2,2H3;3*1H;/q;;;;+3/p-3

InChI key

YMLFYGFCXGNERH-UHFFFAOYSA-K

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

L M Allan et al.
AIHAJ : a journal for the science of occupational and environmental health and safety, 61(6), 820-824 (2001-02-24)
An initial screening test compared the use of tropolone (2-hydroxy-2,4-6-cycloheptatrienone) in acetic acid with sodium diethyldithiocarbamate (NaDDC) as chelating agents for the extraction of butyltin chlorides from glass fiber filters and XAD-2 resin tube. NaDDC was chosen for subsequent analyses.
M Monperrus et al.
Analytical and bioanalytical chemistry, 381(4), 854-862 (2004-12-17)
A robust method has been developed for simultaneous determination of mercury and butyltin compounds in aqueous samples. This method is capable of providing accurate results for analyte concentrations in the picogram per liter to nanogram per liter range. The simultaneous
Dagmar D Doering et al.
Steroids, 67(10), 859-867 (2002-09-17)
Butyltins are widely used biocides and accumulate in the food chain. Tributyltin is an imposex-inducing endocrine disrupter in animals. Imposex is characterized by the development of additional male sex organs on females. In a previous study, we identified tributyltin as
K Senthilkumar et al.
Environmental science and pollution research international, 8(1), 35-47 (2001-05-22)
Concentrations of polychlorinated biphenyls (PCBs), DDT and its metabolites (DDTs), HCH isomers (HCHs), chlordane compounds (CHLs) and hexachlorobenzene (HCB) were determined in sediment, soil, whole body homogenates of resident and migratory birds and their prey items (including fish, green mussel
H Nakata et al.
Environmental pollution (Barking, Essex : 1987), 120(2), 245-253 (2002-10-25)
The effects of exposure to butyltin compounds (BTs: tributyltin; TBT, dibutyltin; DBT and monobutyltin; MBT) and non-ortho coplanar PCBs (IUPAC 77, 126 and 169) on marine mammals and human lymphocyte were evaluated. Peripheral blood mononuclear cells (PBMCs) isolated from Dall's

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service