Skip to Content
Merck
All Photos(3)

Key Documents

197602

Sigma-Aldrich

Quinuclidine

97%

Synonym(s):

1-Azabicyclo[2.2.2]octane, ABCO

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H13N
CAS Number:
Molecular Weight:
111.18
Beilstein:
103111
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

1.5 mmHg ( 20 °C)

Assay

97%

form

solid

mp

157-160 °C (lit.)

solubility

H2O: very slightly soluble
H2O: very soluble
alcohol: miscible
diethyl ether: miscible
organic solvents: very soluble

SMILES string

C1CN2CCC1CC2

InChI

1S/C7H13N/c1-4-8-5-2-7(1)3-6-8/h7H,1-6H2

InChI key

SBYHFKPVCBCYGV-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Quinuclidine is a bicyclic tertiary amine with a bridgehead nitrogen atom structure, often used in the synthesis of iodonium complexes and as a hydrogen-atom-transfer (HAT) catalyst in photo induced reactions. [1]

Application

Quinuclidine can be used as a catalyst:
  • In the Baylis-Hillman reaction of aldehydes with methyl acrylate[2].
  • For the epimerization of α-methylglucose to α-methylallose[3].
It can also be used as a reactant to synthesize 5-[4-[2-(acetyloxy)ethyl]-1-piperidinyl]-2-pyridinecarbonitrile by reacting with 5-bromo-2-pyridinecarbonitrile in the presence of potassium acetate[4].

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 3 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Yong Wang et al.
Nature, 578(7795), 403-408 (2020-01-16)
Glycans have diverse physiological functions, ranging from energy storage and structural integrity to cell signalling and the regulation of intracellular processes1. Although biomass-derived carbohydrates (such as D-glucose, D-xylose and D-galactose) are extracted on commercial scales, and serve as renewable chemical
Journal of the American Chemical Society, 115, 7047-7047 (1993)
H P Kertscher et al.
Die Pharmazie, 46(11), 772-774 (1991-11-01)
A series of 13 PAF-analogues with heterocyclane head groups and variation of the P-N-distance on the C-3-position of the backbone were synthesized. The proaggregatory and inhibitory potencies on rabbit and human blood platelets in vitro was evaluated. Investigations of structure-activity
H P Kertscher et al.
Die Pharmazie, 47(3), 172-174 (1992-03-01)
A series of 11 PAF-analogues, structurally modified in position 1 (alkylcarbamoyloxy), position 2 (n-propyl), and position 3 (polar head group) were synthesized, and the inhibitory potencies on human blood platelets in vitro was evaluated. Investigations of structure-activity relationships revealed, that
Xueling Mi et al.
The Journal of organic chemistry, 70(6), 2338-2341 (2005-03-12)
[reaction: see text] The ionic liquid-bound quinuclidine catalyzed Baylis-Hillman reactions were investigated. The IL-supported catalyst showed equally good catalytic activity as compared with its nonimmobilized counterpart. The corresponding Baylis-Hillman adducts were obtained in moderate to high yields in all the

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service