Skip to Content
Merck
All Photos(1)

Documents

196282

Sigma-Aldrich

3-Thiophenecarboxaldehyde

98%

Synonym(s):

3-Formylthiophene, 3-Thienaldehyde, 3-Thienylcarboxaldehyde, 3-Thiophenealdehyde, 3-Thiophenecarbaldehyde, Thiofuran-3-carboxaldehyde, Thiophen-3-aldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H4OS
CAS Number:
Molecular Weight:
112.15
Beilstein:
105889
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

0.31 mmHg ( 20 °C)

Assay

98%

form

liquid

autoignition temp.

>392 °F

refractive index

n20/D 1.583 (lit.)

bp

194-196 °C (lit.)
86-87 °C/20 mmHg (lit.)

density

1.28 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

[H]C(=O)c1ccsc1

InChI

1S/C5H4OS/c6-3-5-1-2-7-4-5/h1-4H

InChI key

RBIGKSZIQCTIJF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Thiophenecarboxaldehyde has been used in the synthesis of:
  • series of 4-substituted 2-thiophenesulfonamides
  • acetal and ketal derivatives of 4′-demethylepipodophyllotoxin-β-D-glucoside and epipodophyllotoxin-β-D-glucoside
  • 1,2-di-3-thienyl-2-hydroxyethanone (3,3′-thenoin), 3-thienyl symmetric analog of benzoin

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

167.0 °F - closed cup

Flash Point(C)

75 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R S Gupta et al.
Anti-cancer drug design, 2(1), 1-12 (1987-08-01)
We have synthesized acetal and ketal derivatives of 4'-demethylepipodophyllotoxin-beta-D-glucoside (DMEPG) and epipodophyllotoxin-beta-D-glucoside (EPG) with a number of different aldehydes (viz. acetaldehyde, propionaldehyde, 2-thiophenecarboxaldehyde, 3-thiophenecarboxaldehyde, 2-furancarboxaldehyde, benzaldehyde, phenylacetaldehyde, hydrocinnamaldehyde) and acetone. The cross resistance of these compounds towards a set of
J M Holmes et al.
Journal of medicinal chemistry, 37(11), 1646-1651 (1994-05-27)
A series of 4-substituted 2-thiophenesulfonamides was prepared from 3-thiophenecarboxaldehyde using metalation chemistry developed for 3-furaldehyde. Several of these compounds inhibit carbonic anhydrase II in vitro at concentrations of less than 10 nM. In addition, none of these compounds exhibit sensitization
1, 2-Di-3-thienyl-2-hydroxyethanone (3, 3′-thenoin).
Crundwell G, et al.
Acta Crystallographica Section E, Structure Reports Online, 58(6), o668-o670 (2002)
Kollur Shiva Prasad et al.
Molecules (Basel, Switzerland), 25(12) (2020-06-26)
Herein we report the synthesis and structural elucidation of two novel imine-based ligands, 2-(1,10-phenanthrolin-5-yl)imino)methyl)-5-bromophenol (PIB) and N-(1,10-phenanthrolin-5-yl)-1-(thiophen-3-yl)methanimine (PTM) ligands. An in vitro cytotoxicity assay of the synthesized molecules was carried out against breast, cervical, colorectal, and prostate cancer cell lines
Kyungsil Yoon et al.
Cells, 8(3) (2019-03-15)
Chicken ovalbumin upstream promoter-transcription factor I (COUP-TFI) is an orphan receptor and member of the nuclear receptor superfamily. Among a series of methylene substituted diindolylmethanes (C-DIMs) containing substituted phenyl and heteroaromatic groups, we identified 1,1-bis(3'-indolyl)-1-(4-pyridyl)-methane (DIM-C-Pyr-4) as an activator of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service