Skip to Content
Merck
All Photos(1)

Documents

19390

Sigma-Aldrich

Vinyl butyrate

contains 20 ppm 4-methoxyphenol as stabilizer, ≥99.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2COOCH=CH2
CAS Number:
Molecular Weight:
114.14
Beilstein:
1744933
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.0% (GC)

contains

20 ppm 4-methoxyphenol as stabilizer

refractive index

n20/D 1.410

density

0.899 g/mL at 20 °C

storage temp.

2-8°C

SMILES string

CCCC(=O)OC=C

InChI

1S/C6H10O2/c1-3-5-6(7)8-4-2/h4H,2-3,5H2,1H3

InChI key

MEGHWIAOTJPCHQ-UHFFFAOYSA-N

General description

Vinyl butyrate causes the transesterification of natural flavonoid bergenin immobilized onto carboxylic acid functionalized controlled pore glass.

Application

Vinyl butyrate was used as acyl donor in the synthesis of citronellyl esters (acetate, propionate, butyrate, caprate and laurate) using immobilized Candida antarctical ipase B.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

68.0 °F - closed cup

Flash Point(C)

20 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Umar Akbar et al.
Tetrahedron letters, 51(8), 1220-1220 (2010-02-23)
The natural flavonoid bergenin was directly immobilized onto carboxylic acid functionalized controlled pore glass (carboxy-CPG) at 95% yield. Immobilized bergenin was brominated via chloroperoxidase in aqueous solution and then transesterified with vinyl butyrate in diisopropyl ether by subtilisin carslberg (SC)
Betzaida Castillo et al.
BMC biotechnology, 6, 51-51 (2006-12-26)
Enzymes have been extensively used in organic solvents to catalyze a variety of transformations of biological and industrial significance. It has been generally accepted that in dry aprotic organic solvents, enzymes are kinetically trapped in their conformation due to the
Mozaffar Shakeri et al.
Journal of biotechnology, 145(3), 281-283 (2009-12-02)
We examined the activity of free Rhizopus oryzae lipase (ROL) and ROL immobilized on mesoporous materials in transesterification reactions in various dialkylimidazolium-cation based ionic liquids. For free ROL, the highest activity (0.39 U/mg protein) was obtained in [OMIm][PF(6)] followed by
Ming Jian Liu et al.
Drug delivery, 23(9), 3444-3451 (2016-05-21)
The butyryl galactose ester-modified coix component microemulsions (But-Gal-CMEs) was developed for enhanced liver tumor-specific targeting. The study was aimed to evaluate the hepatoma-targeting potential of But-Gal-CMEs in vitro and in vivo. But-Gal-CMEs with a uniform spherical shape exhibited a small
M Holmquist et al.
Journal of protein chemistry, 14(4), 217-224 (1995-05-01)
To reveal the functional role of Glu87 and Trp89 in the lid of Humicola lanuginosa lipase, site-directed mutagenesis at Glu87 and Trp89 was carried out. The catalytic performance of wild-type and mutated lipases was studied in transesterification reactions in cyclohexane

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service