Skip to Content
Merck
All Photos(2)

Key Documents

191434

Sigma-Aldrich

4-Nitrobenzylamine hydrochloride

97%

Synonym(s):

(4-Nitrophenyl)methanamine hydrochloride, 4-Nitrobenzenemethanamine hydrochloride, p-Nitrobenzylamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4CH2NH2 · HCl
CAS Number:
Molecular Weight:
188.61
Beilstein:
3629994
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

~265 °C (dec.) (lit.)

solubility

methanol:glacial acetic acid (1:1): soluble 25 mg/mL, clear, colorless to light yellow

functional group

amine
nitro

SMILES string

Cl.NCc1ccc(cc1)[N+]([O-])=O

InChI

1S/C7H8N2O2.ClH/c8-5-6-1-3-7(4-2-6)9(10)11;/h1-4H,5,8H2;1H

InChI key

SMIXZZMSWYOQPW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

4-Nitrobenzylamine hydrochloride was used in chemical modification of graphite powder and multiwalled carbon nanotubes. It was also used in the preparation of 2-fluoro-6-(4-nitrohenzylamino)purine.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Z Kirkel' et al.
Voprosy meditsinskoi khimii, 32(2), 118-125 (1986-03-01)
Oxidation of p-nitro- and p-dimethylaminomethyl derivatives of benzylamine, catalyzed by amine oxidases from human placenta and blood serum, was studied. The amine oxidase activity was estimated by means of a spectrophotometric procedure involving measurement of aldehyde formed during the reaction
Combinatorial synthesis of 2, 9-substituted purines.
Gray NS, et al.
Tetrahedron Letters, 38(7), 1161-1164 (1997)
Gregory G Wildgoose et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 6(2), 352-362 (2005-03-09)
We demonstrate that graphite powder and multiwalled carbon nanotubes (MWCNTs) can be derivatised by 4-nitrobenzylamine (4-NBA) simply by stirring the graphite powder or MWCNTs in a solution of acetonitrile containing 10 mM 4-NBA. We propose that 4-NBA partially intercalates at
M A Akyüz et al.
Journal of neural transmission (Vienna, Austria : 1996), 114(6), 693-698 (2007-04-03)
Computational studies using the ONIOM methods have been performed to probe the catalytic roles of tyrosine residues 398 and 435 which constitute the "aromatic cage" in the active site of MAO-B. The results presented here provide additional new insights into
The synthesis of a light-switchable amino acid for inclusion into conformationally mobile peptides.
Ulysse L and Chmielewski J.
Bioorganic & Medicinal Chemistry Letters, 4(17), 2145-2146 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service