Skip to Content
Merck
All Photos(1)

Key Documents

18805

Sigma-Aldrich

6-Hydroxyindole

≥99.0% (GC)

Synonym(s):

6-Indolol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7NO
CAS Number:
Molecular Weight:
133.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.0% (GC)

form

solid

mp

126-132 °C

SMILES string

Oc1ccc2cc[nH]c2c1

InChI

1S/C8H7NO/c10-7-2-1-6-3-4-9-8(6)5-7/h1-5,9-10H

InChI key

XAWPKHNOFIWWNZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Reactant for preparation of tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles as potential anticancer immunomodulators
  • Reactant for asymmetrical synthesis of notoamide J as a potential biosynthetic precursor of prenylated indole alkaloids
  • Reactant for preparation of (quinolinyloxymethyl)isoxazolecarboxylate esters antituberculosis agents
  • Reactant for preparation of indolyl(propanolamine) derivatives as HIV inhibitors
  • Reactant for preparation of indoleoxyacetic acid derivatives as peroxisome proliferator-activated receptor agonists
  • Reactant for preparation of 1-aroylindole 3-aroylindoles combretastatin A-4 analogs as antitumor agents and tubulin polymerization inhibitors

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jintae Lee et al.
Applied and environmental microbiology, 73(13), 4100-4109 (2007-05-08)
Since indole is present at up to 500 microM in the stationary phase and is an interspecies biofilm signal (J. Lee, A. Jayaraman, and T. K. Wood, BMC Microbiol. 7:42, 2007), we investigated hydroxyindoles as biofilm signals and found them
Yoshimitsu Yamazaki et al.
Zeitschrift fur Naturforschung. C, Journal of biosciences, 65(1-2), 49-54 (2010-04-02)
A recent study showed that N-acylserotonin derivatives have strong inhibitory activity against tyrosinase. To clarify the role of the 5-hydroxy group in the indole ring, 2-, 4-, 5-, 6-, and 7-hydroxyindole and 11 related compounds such as 5-hydroxyindan and 6-hydroxyquinoline
Stefan W Toennes et al.
Drug testing and analysis, 10(4), 644-650 (2017-10-03)
Each year, synthetic cannabinoids occur in high numbers on the illicit drug market, but data on their detectability are rarely available. A pilot study was performed to assess adverse effects of JWH-018, which is one of the oldest and best
Damon Borg et al.
Journal of analytical toxicology, 41(1), 6-16 (2016-09-30)
Synthetic cannabinoids are a group of psychoactive compounds that mimic the effects of Δ9-tetrahydrocannabinol, the primary psychoactive constituent of marijuana (Cannabis sativa L). The Drug Enforcement Administration has classified many of the most common cannabinoids as Schedule 1 controlled substances.
Nenad Manevski et al.
PloS one, 8(1), e54767-e54767 (2013-02-02)
Human UDP-glucuronosyltransferases (UGTs) are important enzymes in metabolic elimination of endo- and xenobiotics. It was recently shown that addition of fatty acid free bovine serum albumin (BSA) significantly enhances in vitro activities of UGTs, a limiting factor in in vitro-in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service