Skip to Content
Merck
All Photos(2)

Documents

179701

Sigma-Aldrich

Triethylborane solution

1.0 M in THF

Synonym(s):

Triethylboron

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)3B
CAS Number:
Molecular Weight:
97.99
Beilstein:
1731462
MDL number:
UNSPSC Code:
12352001
eCl@ss:
38120201
PubChem Substance ID:
NACRES:
NA.22

form

liquid

reaction suitability

reagent type: reductant

concentration

1.0 M in THF

density

0.865 g/mL at 25 °C

SMILES string

CCB(CC)CC

InChI

1S/C6H15B/c1-4-7(5-2)6-3/h4-6H2,1-3H3

InChI key

LALRXNPLTWZJIJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Catalyst for:
  • Allylation of aldehydes
  • Decarboxylative C-C bond cleavage reactions
  • Rhenium hydride / boron Lewis acid cocatalysis of alkene hydrogenations
  • Regioselective hydroxyalkylation of unsaturated oxime ethers

Reactant for radical reductions of alkyl bromides with N-heterocyclic carbene boranes

Reactant for synthesis of tetramethylammonium trialkylphenylborate salts with oxidation potential
Triethylborane is used as a catalyst for:
  • Allylation of aldehydes
  • Decarboxylative C-C bond cleavage reactions
  • Rhenium hydride / boron Lewis acid cocatalysis of alkene hydrogenations
  • Regioselective hydroxyalkylation of unsaturated oxime ethers


It can be employed as a reactant for radical reductions of alkyl bromides with N-heterocyclic carbene boranes and for the synthesis of tetramethylammonium trialkylphenylborate salts.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Pyr. Liq. 1 - Skin Corr. 1A - STOT SE 3 - Water-react 2

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 1

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rhenium hydride/boron Lewis acid cocatalysis of alkene hydrogenations: Activities comparable to those of precious metal systems.
Jiang, Yanfeng and Hess, Jeannine and Fox, Thomas and Berke, Heinz
Journal of the American Chemical Society, 132(51), 18233-18247 (2010)
Highly enantioselective palladium-catalyzed umpolung allylation of aldehydes.
Zhu, Shou-Fei and Qiao, Xiang-Chen and Zhang, Yong-Zhen and Wang, Li-Xin and Zhou, Qi-Lin
Chemical Science, 2(6), 1135-1140 (2011)
Synthesis of N-heterocyclic carbene boranes via silver N-heterocyclic carbene complexes.
Ono S, et al.
Polyhedron, 137(20), 296-305 (2017)
Aerobic Hydroxylation of N-Borylenamine: Triethylborane-Mediated Hydroxyalkylation of α, β-Unsaturated Oxime Ether.
Ueda M, et al.
Organic Letters, 11(20), 4632-4635 (2009)
Formation of Ketenimines via the Palladium-Catalyzed Decarboxylative π-Allylic Rearrangement of N-Alloc Ynamides.
Alexander, Juliana R and Cook, Matthew J
Organic Letters, 19(21), 5822-5825 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service