179000
Benzo-1,4-dioxane
97%
Synonym(s):
1,2-Ethylenedioxybenzene, 1,4-Benzodioxan, 2,3-Dihydro-1,4-benzodioxin
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C8H8O2
CAS Number:
Molecular Weight:
136.15
Beilstein:
120846
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
Recommended Products
Assay
97%
form
liquid
refractive index
n20/D 1.549 (lit.)
bp
103 °C/6 mmHg (lit.)
density
1.142 g/mL at 25 °C (lit.)
SMILES string
C1COc2ccccc2O1
InChI
1S/C8H8O2/c1-2-4-8-7(3-1)9-5-6-10-8/h1-4H,5-6H2
InChI key
BNBQRQQYDMDJAH-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
The anti-inflammamtory properties and synthesis of carboxylic acid compounds containing benzo-1,4-dioxane (2,3-dihydro-1,4-benzodioxin) subunit was studied.
Application
Benzo-1,4-dioxane (2,3-dihydro-1,4-benzodioxin) was used in the synthesis of stereoisomers which were evaluated as α- and β- adrenergic antagonists.
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
156.2 °F - closed cup
Flash Point(C)
69 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
M Khouili et al.
Farmaco (Societa chimica italiana : 1989), 51(3), 185-188 (1996-03-01)
The four stereoisomers of compound 1 were synthesized from 2,3-dihydro-1, 4-benzodioxin and evaluated as alpha- and beta-adrenergic antagonists. Enantiomer 1-b [2R, 2'S] (Figure 1) is the best beta 1-blocking agent. Furthermore all compounds showed a alpha-blocking activity.
M T Vázquez et al.
Farmaco (Societa chimica italiana : 1989), 51(3), 215-217 (1996-03-01)
Synthesis and antiinflammatory properties of new carboxylic acids containing the 2,3-dihydro-1,4-benzodioxin subunit are described. The 2-(2,3-dihydro-1, 4-benzodioxin-6-yl)acetic acid was of comparable potency to Ibuprofen, in carrageenan induced rat paw edema assay.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service