177458
4-Acetylbenzoic acid
98%
Synonym(s):
Acetophenone-4-carboxylic acid
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
CH3COC6H4CO2H
CAS Number:
Molecular Weight:
164.16
Beilstein:
2207355
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
98%
mp
208-210 °C (lit.)
functional group
carboxylic acid
ketone
SMILES string
CC(=O)c1ccc(cc1)C(O)=O
InChI
1S/C9H8O3/c1-6(10)7-2-4-8(5-3-7)9(11)12/h2-5H,1H3,(H,11,12)
InChI key
QBHDSQZASIBAAI-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
4-Acetylbenzoic acid was used in the preparation of an ester-derivative of paclitaxel.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Adam W G Alani et al.
Biomaterials, 31(7), 1765-1772 (2009-12-05)
Poly(ethylene glycol)-block-poly(aspartate-hydrazide) (PEG-p(Asp-Hyd)) was modified using either levulinic acid (LEV) or 4-acetyl benzoic acid (4AB) attached via hydrazone bonds. Paclitaxel (PTX) conjugated to the linkers formed PEG-p(Asp-Hyd-LEV-PTX) and PEG-p(Asp-Hyd-4AB-PTX). PEG-p(Asp-Hyd-LEV-PTX) and PEG-p(Asp-Hyd-4AB-PTX) assemble into unimodal polymeric micelles with diameters of
Yamin Wang et al.
Journal of pharmaceutical and biomedical analysis, 46(1), 66-74 (2007-10-19)
The similarities and differences of essential oil components in Pericarpium Citri Reticulatae Viride (PCRV) and Pericarpium Citri Reticulatae (PCR) were investigated by GC-MS combined with a chemometric method, named alternative moving window factor analysis (AMWFA). Furthermore, temperature-programmed retention indices (PTRIs)
Serge Pauillac et al.
Journal of immunological methods, 263(1-2), 75-83 (2002-05-16)
We describe here a protocol for determining the epitope density of hapten-carrier conjugates at the nanomolar level. Conjugates of bovine serum albumin (BSA) and hen egg albumin (OVA) were prepared with two model haptens: 4-acetyl benzoic acid (ABA) as a
Synthesis and in vitro efficacy of acid-sensitive poly(ethylene glycol) paclitaxel conjugates.
Paula Rodrigues et al.
Bioorganic & medicinal chemistry letters, 13(3), 355-360 (2003-07-29)
Carina I C Crucho et al.
Materials science & engineering. C, Materials for biological applications, 109, 110528-110528 (2020-04-02)
Thermally activated delayed fluorescence (TADF) has revolutionized the field of organic light emitting diodes owing to the possibility of harvesting non-emissive triplet states and converting them in emissive singlet states. This mechanism generates a long-lived delayed fluorescence component which can
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service