Skip to Content
Merck
All Photos(1)

Documents

176990

Sigma-Aldrich

tert-Amylamine

98%

Synonym(s):

1,1-Dimethylpropylamine, tert-Pentylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5C(CH3)2NH2
CAS Number:
Molecular Weight:
87.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.3996 (lit.)

bp

77 °C (lit.)

density

0.746 g/mL at 25 °C (lit.)

SMILES string

CCC(C)(C)N

InChI

1S/C5H13N/c1-4-5(2,3)6/h4,6H2,1-3H3

InChI key

GELMWIVBBPAMIO-UHFFFAOYSA-N

Related Categories

Application

tert-Amylamine was used as a substitute for t-butylamino substituent in cyanobenzylamine derivative.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

30.2 °F - closed cup

Flash Point(C)

-1 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Afrooz Shiekhzadeh et al.
Applied biochemistry and biotechnology, 190(2), 506-528 (2019-08-08)
In this paper, a new anticancer Pt (II) complex, cis-[Pt (NH3)2(tertpentylgly)]NO3, was synthesized with glycine-derivative ligand and characterized. Cytotoxicity of this water-soluble Pt complex was studied against human cancer breast cell line of MCF-7. The interaction of human serum albumin
A M Gilbert et al.
Journal of medicinal chemistry, 43(6), 1203-1214 (2000-03-29)
A novel series of benzylamine, potassium channel openers (KCOs) is presented as part of our program toward designing new, bladder-selective compounds for the treatment of urge urinary incontinence (UUI). We have found that the in vitro potency of (R)-4-[3,4-dioxo-2-(1,2, 2-trimethyl-propylamino)-cyclobut-1-enylamino]-3-ethyl-benzo
Ting Su et al.
Analytical biochemistry, 597, 113654-113654 (2020-03-07)
Sepsis is the leading cause of death in hospitalized patients and is characterized by a dysregulated inflammatory response to infection and multiple organ failure, including the liver. Transglutaminase 2 (TG2) is a multifunctional enzyme that exhibits transamidase, GTPase, and integrin-binding
Soudeh Hosseinzadeh et al.
Journal of biomolecular structure & dynamics, 38(8), 2215-2228 (2019-06-18)
Cisplatin, carboplatin and oxaliplatin and their analogs are effective anticancer agents, but their clinical using is limited by some serious side effects. S,S donor ligands such as dithiocarbamates can be used to reduce some side effects. In this study, some
Radhe Shyam et al.
Biopolymers, 110(6), e23273-e23273 (2019-03-22)
The design of linear peptoid oligomers adopting well-defined secondary structures while mimicking defined peptide primary sequences is a major challenge in the context of drug discovery. To this end, chemists have developed cis-inducing peptoid side chains to build robust polyproline

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service