Skip to Content
Merck
All Photos(2)

Key Documents

165794

Sigma-Aldrich

3-Aminobenzenesulfonic acid

97%

Synonym(s):

Metanilic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H4SO3H
CAS Number:
Molecular Weight:
173.19
Beilstein:
473264
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

97%

form

powder

mp

>300 °C (lit.)

solubility

ethanol: very slightly soluble(lit.)
methanol: very slightly soluble(lit.)

functional group

sulfonic acid

SMILES string

Nc1cccc(c1)S(O)(=O)=O

InChI

1S/C6H7NO3S/c7-5-2-1-3-6(4-5)11(8,9)10/h1-4H,7H2,(H,8,9,10)

InChI key

ZAJAQTYSTDTMCU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Aminobenzenesulfonic acid undergoes microbial desulfonation to 3-aminophenol by Pseudomonas sp. strain S-313[1].

Application

3-Aminobenzenesulfonic acid was used in the synthesis of cytocompatible sulfonated polyanilines[2]. It was used in fabrication of novel glucose biosensor having large active surface area and excellent conductivity[3].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simon Doswald et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 27(12), 4108-4114 (2020-12-23)
The functionalization of magnetic nanoparticles has been an important field in the last decade due to the versatile applications in catalysis and biomedicine. Generally, a high degree of functionalities on the surface of the nanoparticles is desired. In this study
D Zürrer et al.
Applied and environmental microbiology, 53(7), 1459-1463 (1987-07-01)
Sulfur-limited batch enrichment cultures containing one of nine multisubstituted naphthalenesulfonates and an inoculum from sewage yielded several taxa of bacteria which could quantitatively utilize 19 sulfonated aromatic compounds as the sole sulfur source for growth. Growth yields were about 4
Yanyin Yang et al.
Macromolecular rapid communications, 32(12), 887-892 (2011-05-19)
We report here that by good design, polyaniline (PANI) can be cytocompatible and formed into usable scaffolds for bio-medical applications. By adjusting the ratio of two monomers, aniline (AN) and metanilic acid (MA), a series of copolymers with different sulfonation
Mei-Hwa Lee et al.
Biosensors & bioelectronics, 150, 111901-111901 (2019-11-27)
Molecularly imprinted polymers (MIPs) have been developed to replace antibodies for the recognition of target molecules (such as antigens), and have been integrated into electrochemical sensing approaches by polymerization onto an electrode. Electrochemical sensing is inexpensive and flexible, and has
Kai-Hsi Liu et al.
Biosensors, 10(3) (2020-03-04)
Molecularly imprinted polymers (MIPs) can often bind target molecules with high selectivity and specificity. When used as MIPs, conductive polymers may have unique binding capabilities; they often contain aromatic rings and functional groups, which can undergo π-π and hydrogen bonding

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service