Skip to Content
Merck
All Photos(1)

Key Documents

162973

Sigma-Aldrich

2-[2-(2-Chloroethoxy)ethoxy]ethanol

96%

Synonym(s):

Triethylene glycol monochlorohydrin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl(CH2CH2O)2CH2CH2OH
CAS Number:
Molecular Weight:
168.62
Beilstein:
1739194
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

form

liquid

refractive index

n20/D 1.458 (lit.)

bp

117-120 °C/5 mmHg (lit.)

density

1.16 g/mL at 25 °C (lit.)

SMILES string

OCCOCCOCCCl

InChI

1S/C6H13ClO3/c7-1-3-9-5-6-10-4-2-8/h8H,1-6H2

InChI key

KECMLGZOQMJIBM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-[2-(2-Chloroethoxy)ethoxy]ethanol was used in the synthesis of 1-[3-(2-aminoethyl)-1H-indol-5-yloxy]-3,6-dioxa-8-mercaptooctane. It was also used in the synthesis of modified oligonucleotides bearing a disulfide or a thiol function at the 3′-end and other modifications at the 5′-end.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

224.6 °F - closed cup

Flash Point(C)

107 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sandra J Rosenthal et al.
Journal of the American Chemical Society, 124(17), 4586-4594 (2002-04-25)
To explore the potential for use of ligand-conjugated nanocrystals to target cell surface receptors, ion channels, and transporters, we explored the ability of serotonin-labeled CdSe nanocrystals (SNACs) to interact with antidepressant-sensitive, human and Drosophila serotonin transporters (hSERT, dSERT) expressed in
Po-Chih Yang et al.
Polymers, 11(12) (2019-12-11)
This paper gathered studies on multistimulus-responsive sensing and self-assembly behavior of a novel amphiphilic diblock copolymer through a two-step reverse addition-fragmentation transfer (RAFT) polymerization technique. N-Isopropylacrylamide (NIPAM) macromolecular chain transfer agent and diblock copolymer (poly(NIPAM-b-Azo)) were discovered to have moderate
Solid phase synthesis of 5', 3'-bifunctional oligodeoxyribonucleotides bearing a masked thiol group at the 3'-end.
Bonfils E and Thuong NT.
Tetrahedron Letters, 32(26), 3053-3056 (1991)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service