Skip to Content
Merck
All Photos(1)

Documents

162353

Sigma-Aldrich

3-Methyl-2-buten-1-ol

99%

Synonym(s):

3,3-Dimethylallyl alcohol, Prenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=CHCH2OH
CAS Number:
Molecular Weight:
86.13
Beilstein:
1633479
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39020334
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

1.4 mmHg ( 20 °C)

Assay

99%

form

liquid

expl. lim.

16.3 %

refractive index

n20/D 1.443 (lit.)

bp

140 °C (lit.)

density

0.848 g/mL at 25 °C (lit.)

SMILES string

C\C(C)=C\CO

InChI

1S/C5H10O/c1-5(2)3-4-6/h3,6H,4H2,1-2H3

InChI key

ASUAYTHWZCLXAN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Methyl-2-buten-1-ol reacts with nitrosocarbonyl benzene to yield 5-hydroxy-isoxazolidines. It is commonly used as fragrance ingredient.

Application

3-Methyl-2-buten-1-ol was used as starting reagent during asymmetric total syntheses of (R)-(+)- and (S)-(-)-umbelactone via Sharpless asymmetric epoxidation reaction.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

124.7 °F - closed cup

Flash Point(C)

51.5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anastasia Zerva et al.
Molecules (Basel, Switzerland), 23(9) (2018-09-22)
Feruloyl esterases (FAEs, E.C. 3.1.1.73) are biotechnologically important enzymes with several applications in ferulic acid production from biomass, but also in synthesis of hydroxycinnamic acid derivatives. The use of such biocatalysts in commercial processes can become feasible by their immobilization
Charla A Centrone et al.
Bioorganic & medicinal chemistry, 12(21), 5495-5503 (2004-10-07)
Mycobacteria biosynthesize a cell wall structure that is rich in polysaccharides containing arabinofuranose residues. The source of these arabinofuranose residues is decaprenolphosphoarabinose (1), the donor substrate for mycobacterial arabinosyltransferases. We have previously demonstrated that an analog of 1, C-phosphonate 7
Dusan Hesek et al.
Journal of the American Chemical Society, 134(33), 13881-13888 (2012-08-07)
We describe a practical, multigram synthesis of (2Z,6Z,10Z,14Z,18E,22E)-3,7,11,15,19,23,27-heptamethyl-2,6,10,14,18,22,26-octacosaheptaen-1-ol [(Z(4),E(2),ω)-heptaprenol, 4] using the nerol-derived sulfone 8 as the key intermediate. Sulfone 8 is prepared by the literature route and is converted in five additional steps (18% yield from 8) to (Z(4),E(2),ω)-heptaprenol
Teris A van Beek
Journal of chromatography. A, 967(1), 21-55 (2002-09-11)
The chemical analysis and quality control of Ginkgo leaves and extracts is reviewed. Important constituents present in the medicinally used leaves are the terpene trilactones, i.e., ginkgolides A, B, C, J and bilobalide, many flavonol glycosides, biflavones, proanthocyanidins, alkylphenols, simple
Alvaro Acosta-Serrano et al.
Eukaryotic cell, 3(2), 255-263 (2004-04-13)
Concanavalin A (ConA) kills the procyclic (insect) form of Trypanosoma brucei by binding to its major surface glycoprotein, procyclin. We previously isolated a mutant cell line, ConA 1-1, that is less agglutinated and more resistant to ConA killing than are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service