161756
Bromomethyl methyl ether
technical grade, 90%
Synonym(s):
1-Bromomethyl methyl ether, Bromomethoxymethane, Methoxymethyl bromide
About This Item
Recommended Products
grade
technical grade
Assay
90%
form
liquid
refractive index
n20/D 1.456 (lit.)
bp
87 °C (lit.)
density
1.531 g/mL at 25 °C (lit.)
storage temp.
2-8°C
SMILES string
COCBr
InChI
1S/C2H5BrO/c1-4-2-3/h2H2,1H3
InChI key
JAMFGQBENKSWOF-UHFFFAOYSA-N
Application
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
80.6 °F - closed cup
Flash Point(C)
27 °C - closed cup
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service