All Photos(1)
About This Item
Linear Formula:
(CH3O)2C6H3COCl
CAS Number:
Molecular Weight:
200.62
Beilstein:
511839
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
form
solid
bp
157-158 °C/16 mmHg (lit.)
mp
43-46 °C (lit.)
functional group
acyl chloride
SMILES string
COc1cc(OC)cc(c1)C(Cl)=O
InChI
1S/C9H9ClO3/c1-12-7-3-6(9(10)11)4-8(5-7)13-2/h3-5H,1-2H3
InChI key
FTHPLWDYWAKYCY-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
3,5-Dimethoxybenzoyl chloride undergoes addition reaction with 4,4-dimethyl-2-pentyne in presence of AlCl3 via 1,2- methyl shift.
Application
3,5-Dimethoxybenzoyl chloride was used to study the mechanism and kinetics of solvolysis of 3,4- and 3,5-dimethoxybenzoyl chlorides in various binary solvents.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
230.0 °F - closed cup
Flash Point(C)
110 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Stereochemistry and mechanism of acylation of acetylenes.
Martens H, et al.
Tetrahedron, 31(2), 177-183 (1975)
Kinetic and Theoretical Consideration of 3, 4-and 3, 5-Dimethoxybenzoyl Chlorides Solvolyses.
Park K-H and Kevill DN.
Bull. Korean Chem. Soc., 34(10), 2989-2989 (2013)
Dongsheng Xu et al.
Journal of chromatography. A, 1593, 63-72 (2019-02-05)
An O-[2-(methacryloyloxy)-ethylcarbamoyl]-10,11-dihydroquinidine (MQD)-silica hybrid monolithic column was prepared by a facile "one-step" strategy within a 100 μm I.D. capillary. The influence of the methanol, ethylene glycol and water volume ratio, reaction temperature and time, cetyltrimethylammonium bromide and MQD monomers content
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service