Skip to Content
Merck
All Photos(2)

Key Documents

161497

Sigma-Aldrich

3,4-(Methylenedioxy)aniline

97%

Synonym(s):

5-Amino-1,3-benzodioxole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7NO2
CAS Number:
Molecular Weight:
137.14
Beilstein:
4919
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

97%

bp

144 °C/16 mmHg (lit.)

mp

39-41 °C (lit.)

SMILES string

Nc1ccc2OCOc2c1

InChI

1S/C7H7NO2/c8-5-1-2-6-7(3-5)10-4-9-6/h1-3H,4,8H2

InChI key

XGNXYCFREOZBOL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3,4-(Methylenedioxy)aniline is an pharmaceutically important aniline derivative[1]. It undergoes N-alkylation with cyclic secondary alkylamines in the presence of Shvo catalyst to yield N-arylpyrrolidines[1].

Application

3,4-(Methylenedioxy)aniline was used in the synthesis of γ-glutamylanilides[2].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A general ruthenium-catalyzed synthesis of aromatic amines.
Dirk Hollmann et al.
Angewandte Chemie (International ed. in English), 46(43), 8291-8294 (2007-09-25)
A Rsoswsky et al.
Journal of medicinal chemistry, 22(9), 1034-1037 (1979-09-01)
Nine heretofore unknown mono- and dihydroxyanilide analogues of the cytotoxic mushroom metabolites L-glutamic acid gamma-(4-hydroxyanilide) (1) and L-glutamic acid gamma-(3,4-dihydroxyanilide) (3, agaridoxin) were synthesized and tested as inhibitors of the growth of B16 mouse melanoma cells in culture. The naturally
K N Vennila et al.
Bioorganic chemistry, 81, 184-190 (2018-08-24)
The induced fit docking of anilino quinoline scaffold results in the required hydrogen bonding interactions with amino acid residues in the orthosteric site of 3 Phosphoinositide dependent kinase (PDK1). The rational design of 4-substituted amino quinolines is carried out and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service