Skip to Content
Merck
All Photos(1)

Key Documents

157880

Sigma-Aldrich

S-(Thiobenzoyl)thioglycolic acid

99%

Synonym(s):

(Thiobenzoylthio)acetic acid, Dithiobenzoic acid carboxymethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CS2CH2CO2H
CAS Number:
Molecular Weight:
212.29
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

mp

125-127 °C (lit.)

solubility

acetone: soluble 25 mg/mL, clear

functional group

carboxylic acid
phenyl
thioether

SMILES string

OC(=O)CSC(=S)c1ccccc1

InChI

1S/C9H8O2S2/c10-8(11)6-13-9(12)7-4-2-1-3-5-7/h1-5H,6H2,(H,10,11)

InChI key

XBEIANFIOZTEDE-UHFFFAOYSA-N

Related Categories

General description

S-(Thiobenzoyl)thioglycolic acid (TBTGA) also known as (thiobenzoylthio)acetic acid is a reagent used for thiobenzoylation. It participates in the transformation of threonine into β-methylcysteine.

Application

S-(Thiobenzoyl)thioglycolic acid ((thiobenzoylthio)acetic acid) was employed as reversible addition-fragmentation chain transfer (RAFT) agent during the copolymerization of styrene and divinylbenzene. It was also employed as chain-transfer agent for the RAFT polymerizations of styrene, methyl methacrylate and butyl acrylate.
It may be used to synthesize the following:
  • N-Thiobenzoyl-DL-threonine ethyl ester by reacting with DL-threonine.
  • Thiobenzoic acid O-esters by reacting with alkoxides.
  • N-Thiobenzoylamino acids by reacting with amino acids.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

(Thiocarbonyl-a-thio) carboxylic acid derivatives as transfer agents in reversible addition-fragmentation chain-transfer polymerizations.
Farmer SC and Patten TE.
Journal of Polymer Science Part A: Polymer Chemistry, 40(4), 555-563 (2002)
Copolymerization Kinetics of Styrene and Divinylbenzene in the Presence of S-Thiobenzoyl Thioglycolic Acid as RAFT Agent.
Roa-Luna M, et al.
Chemical Engineering & Technology, 33(11), 1893-1899 (2000)
Thiobenzoylation method of protein sequencing: gas chromatography/mass spectrometric detection of 5-acetoxy-2-phenylthiazoles.
Stolowitz ML, et al.
Methods in Protein Sequence Analysis, 37-44 (1993)
Photochemical transformations. Part XXXI. Photolysis of thiobenzoic acid O-esters. Part II. General methods for the preparation of thiobenzoic acid O-esters.
Barton DHR, et al.
Journal of the Chemical Society. Perkin Transactions 1, 1571-1574 (1973)
Imahori K and Sakiyama F.
Methods in Protein Sequence Analysis, 42-44 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service