Skip to Content
Merck
All Photos(1)

Key Documents

155233

Sigma-Aldrich

3-Piperidinemethanol

96%

Synonym(s):

3-(Hydroxymethyl)piperidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13NO
CAS Number:
Molecular Weight:
115.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
solid
Assay:
96%

Quality Level

Assay

96%

form

solid

bp

106-107 °C/3.5 mmHg (lit.)

density

1.026 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

OCC1CCCNC1

InChI

1S/C6H13NO/c8-5-6-2-1-3-7-4-6/h6-8H,1-5H2

InChI key

VUNPWIPIOOMCPT-UHFFFAOYSA-N

General description

The standard molar energy of combustion of 3-Piperidinemethanol has been studied.

Application

Reactant for synthesis of:
CB2 receptor agonists for the treatment of chronic pain
Spiroimidazolidinone NPC1L1 inhibitors
Quorum sensing modulators
Protein lysine methyltransferase G9a inhibitors
P2Y12 antagonists for inhibition of platelet aggregation
Soluble epoxide hydrolase inhibitors

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manuel A V Ribeiro da Silva et al.
The journal of physical chemistry. B, 109(27), 13356-13362 (2006-07-21)
The standard (p degrees = 0.1 MPa) molar enthalpies of formation of 2-, 3-, and 4-chloroaniline were derived from the standard molar energies of combustion, in oxygen, at T = 298.15 K, measured by rotating bomb combustion calorimetry. The Calvet

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service