Skip to Content
Merck
All Photos(1)

Key Documents

15456

Sigma-Aldrich

Boc-Lys-OH

≥99.0% (NT)

Synonym(s):

Nα-(tert-Butoxycarbonyl)-L-lysine, Nα-Boc-L-lysine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2(CH2)4CH(COOH)NHCOOC(CH3)3
CAS Number:
Molecular Weight:
246.30
Beilstein:
4252546
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥99.0% (NT)

form

solid

optical activity

[α]20/D +4.6±0.5°, c = 2% in H2O

reaction suitability

reaction type: Boc solid-phase peptide synthesis

mp

~205 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

CC(C)(C)OC(=O)N[C@@H](CCCCN)C(O)=O

InChI

1S/C11H22N2O4/c1-11(2,3)17-10(16)13-8(9(14)15)6-4-5-7-12/h8H,4-7,12H2,1-3H3,(H,13,16)(H,14,15)/t8-/m0/s1

InChI key

DQUHYEDEGRNAFO-QMMMGPOBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Boc-Lys-OH also known as Nα-Boc-L-lysine, is an amino acid derivative of lysine which is used in solid phase peptide synthesis.

Application

Boc-Lys-OH is used as a building block to form peptides that have bioorthogonal reactive groups via boc protected solid phase synthesis. [1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Building Blocks for the Construction of Bioorthogonally Reactive Peptides via Solid-Phase Peptide Synthesis
BM Zeglis
ChemistryOpen, 3, 48-53 (2014)
Mingqian Tan et al.
Pharmaceutical research, 31(6), 1469-1476 (2013-03-09)
To synthesize and evaluate a peptide targeted nanoglobular dual modal imaging agent specific to a cancer biomarker in tumor stroma for MRI and fluorescence visualization of prostate tumor in image-guided surgery. A peptide (CGLIIQKNEC, CLT1) targeted generation 2 nanoglobular (polylysine
Katherine A Miller et al.
Journal of bacteriology, 197(17), 2831-2839 (2015-06-24)
Salmonella enteric serovar Typhimurium, a major cause of food-borne illness, is capable of using a variety of carbon and nitrogen sources. Fructoselysine and glucoselysine are Maillard reaction products formed by the reaction of glucose or fructose, respectively, with the ε-amine
Griet Van Zeebroeck et al.
Nature chemical biology, 5(1), 45-52 (2008-12-09)
Transporter-related nutrient sensors, called transceptors, mediate nutrient activation of signaling pathways through the plasma membrane. The mechanism of action of transporting and nontransporting transceptors is unknown. We have screened 319 amino acid analogs to identify compounds that act on Gap1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service