Skip to Content
Merck
All Photos(1)

Key Documents

15149

Sigma-Aldrich

N,N-Bis(phosphonomethyl)glycine

≥98.0% (T)

Synonym(s):

N-(Carboxymethyl)iminodi(methylphosphinic acid), Glyphosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H11NO8P2
CAS Number:
Molecular Weight:
263.08
Beilstein:
1884944
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (T)

functional group

amine
carboxylic acid

SMILES string

O=P(CN(CP(O)(O)=O)CC(O)=O)(O)O

InChI

1S/C4H11NO8P2/c6-4(7)1-5(2-14(8,9)10)3-15(11,12)13/h1-3H2,(H,6,7)(H2,8,9,10)(H2,11,12,13)

InChI key

OXHDYFKENBXUEM-UHFFFAOYSA-N

General description

N,N-Bis(phosphonomethyl)glycine was present in the ligand bound to human apotransferrin.

Application

Application: Complexing agent; pK-values: pK1: ·2, pK2: 2.0, pK3: 5.20, pK4: 6.77, pK5: 10.89

Anti-diabetic; delay onset of diabetes in non-obese NOD mice

Glyphosine binds to a pocket of MHC class II molecules I-Ag7 and DQ8, and modify T cell responses to the autoantigenic insulin B chain fragment 9–23 (B:9–23) peptide. Glyphosine stimulate IL-10 production and delay onset of diabetes in non-obese NOD mice.†

Biochem/physiol Actions

Anti-diabetic; delay onset of diabetes in non-obese NOD mice

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aurelia Visa et al.
Nanomaterials (Basel, Switzerland), 10(5) (2020-05-14)
The rapid increase of industrial activities leads to serious environmental pollution, especially, in aqueous systems and particularly with heavy metals. Cadmium, one of the most poisonous elements, is rapidly accumulated in the human body, therefore, the efficient removal of cadmium
Eva Orejuela et al.
Electrophoresis, 26(23), 4478-4485 (2005-11-01)
A straightforward and sensitive method has been developed for the analysis of phosphorus-containing amino acid herbicides (glufosinate and aminomethylphosphonic acid, the major metabolite of glyphosate) in soil samples. For this purpose, the analytical features of two indocyanine fluorescent dyes, sulfoindocyanine
W R Harris et al.
Biochemistry, 30(28), 6930-6936 (1991-07-16)
Difference ultraviolet spectroscopy has been used to monitor the binding of a series of phosphonate ligands to human apotransferrin. The ligands consist of pyrophosphate as well as the phosphonic acids (aminomethyl)phosphonic acid (AMPA), (hydroxymethyl)phosphonic acid (HMP), (phosphonomethyl)-iminodiacetic acid (PIDA), N,N-bis(phosphonomethyl)glycine
L J Olson et al.
Toxicology, 30(2), 103-114 (1984-03-01)
Exposure to the plant growth regulators, chlorocholine chloride (CCC) and glyphosine (GPS), resulted in significant immunomodulatory effects after feeding to deer mice (Peromyscus maniculatus) for 28 days. Cyclophosphamide (CP) and saline controls were included. Both CCC and GPS feeding levels
Markus Galanski et al.
Journal of medicinal chemistry, 46(23), 4946-4951 (2003-10-31)
A series of osteotropic (bone-seeking) [(bis(phosphonomethyl)amino-kappaN)acetato-kappaO(2-)]platinum(II) complexes attached to diammine, ethane-1,2-diamine, cis-R,S-cyclohexane-1,2-diamine, trans-S,S-cyclohexane-1,2-diamine, or trans-R,R-cyclohexane-1,2-diamine has been synthesized in accord with the concept of drug targeting and characterized by elemental analysis, (1)H, (13)C, and (31)P NMR spectroscopy. The in vitro

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service