Skip to Content
Merck
All Photos(2)

Key Documents

151157

Sigma-Aldrich

Cholesteryl oleyl carbonate

Synonym(s):

3β-Hydroxy-5-cholestene 3-oleyl carbonate, 5-Cholesten-3β-ol 3-oleyl carbonate, 5-Cholesten-3-yl 9-octadecenyl carbonate, Cholesterol 9-octadecenyl carbonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H80O3
CAS Number:
Molecular Weight:
681.13
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

form

liquid crystal

optical activity

[α]27/D −22°, c = 2 in chloroform

SMILES string

CCCCCCCC\C=C/CCCCCCCCOC(=O)O[C@H]1CC[C@]2(C)[C@H]3CC[C@]4(C)[C@H](CC[C@H]4[C@@H]3CC=C2C1)[C@H](C)CCCC(C)C

InChI

1S/C46H80O3/c1-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-34-48-44(47)49-39-30-32-45(5)38(35-39)26-27-40-42-29-28-41(37(4)25-23-24-36(2)3)46(42,6)33-31-43(40)45/h14-15,26,36-37,39-43H,7-13,16-25,27-35H2,1-6H3/b15-14-/t37-,39+,40+,41-,42+,43+,45+,46-/m1/s1

InChI key

XMPIMLRYNVGZIA-TZOMHRFMSA-N

General description

Cholesteryl oleyl carbonate(COC) is a carbonate ester of cholesterol and oleyl alcohol with carbonic acid that is used as a liquid crystal(LC) molecule. It has is a crystalline material with a helical structure.[1][2]

Application

COC can be incorporated in the polyethylene glycol(PEG)-polyurethane(PU) matrix that exhibits lower thrombogenicity and an improved haemocompatibility which can used as a biomaterial in biomedical applications.[3] It may also be self-assembled with cadmium selenide(CdSe) quantum dots(QDs) for the fabrication of novel multifunctional switchable devices.[4]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and evaluation of poly (hexamethylene-urethane) and PEG-poly (hexamethylene-urethane) and their cholesteryl oleyl carbonate composites for human blood biocompatibility.
Shih MF, et al.
Molecules (Basel), 16(10), 8181-8197 (2011)
Viscosity variation of Cholesteryl oleyl carbonate at various temperature with Acoustic parametres.
Singh, Jatinder Pal, and Rajesh Sharma
IOSR Journal of Applied Physics, 3, 88-91 (2013)
Quantum dot self-assembly in liquid crystal media.
Emerging Liquid Crystal Technologies V, 7618(10), 76180F-76180F (2010)
Thermochromic composite fibres containing liquid crystals formed via melt extrusion.
van der Werff L, et al.
J. Mater. Sci., 48(14), 5005-5011 (2013)
Anomalous selective reflection in cholesteryl oleyl carbonate-nematic 5CB mixtures and effects of their doping by single-walled carbon nanotubes
Lebovka, N. I., et al.
Liq. Cryst., 40(7), 968-975 (2013)

Articles

Chiral nematic liquid crystal (N*-LC) facilitates asymmetric acetylene polymerization, yielding helical chains and fibrils in polyacetylene film.

Chiral nematic liquid crystal (N*-LC) facilitates asymmetric acetylene polymerization, yielding helical chains and fibrils in polyacetylene film.

Chiral nematic liquid crystal (N*-LC) facilitates asymmetric acetylene polymerization, yielding helical chains and fibrils in polyacetylene film.

Chiral nematic liquid crystal (N*-LC) facilitates asymmetric acetylene polymerization, yielding helical chains and fibrils in polyacetylene film.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service