All Photos(1)
About This Item
Linear Formula:
[(CH3)3C]2C6H2-2-(OH)CO2H
CAS Number:
Molecular Weight:
250.33
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
mp
157-162 °C (lit.)
functional group
carboxylic acid
SMILES string
CC(C)(C)c1cc(C(O)=O)c(O)c(c1)C(C)(C)C
InChI
1S/C15H22O3/c1-14(2,3)9-7-10(13(17)18)12(16)11(8-9)15(4,5)6/h7-8,16H,1-6H3,(H,17,18)
InChI key
ZWQBZEFLFSFEOS-UHFFFAOYSA-N
Related Categories
Application
3,5-Di-tert-butylsalicylic acid was used to study long wavelength fluorescence emission of 3,5-Di-tert-butylsalicylic acid in a variety of organic solvents. It was also used to catalyze the reaction between aldehydes and silyl ketene acetals.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
354.2 °F - closed cup
Flash Point(C)
179 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Photoinduced proton transfers in 3, 5-di-tert-butylsalicylic acid.
The Journal of Physical Chemistry, 99(32), 12103-12108 (1995)
Veli T Kasumov et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 107, 31-38 (2013-02-19)
A series of new polyfluorinated palladium(II) complexes (7-12) of N-polyfluorophenyl-3,5-di-tert-butylsalicylaldimines (1-6) have been synthesized. They were characterized by analytical, spectroscopic (UV/Vis, IR, (1)H NMR, and ESR), electrochemical methods and their chemical oxidation and hydrogenation catalytic activity were studied. The X-ray
A first example of macromolecular Ti (IV) Lewis acid in the catalytic enantioselective Mukaiyama reaction.
Tetrahedron Asymmetry, 9(9), 1479-1482 (1998)
M V Chidambaram et al.
Journal of pharmaceutical sciences, 80(8), 810-811 (1991-08-01)
The initial yield of 3,5-di-t-butylsalicylic acid obtained via Kolbe-Schmitt carboxylation of the potassium salt of 2,4-di-t-butylphenol was less than 1% and was accompanied by a 65% yield of 2,2'-dihydroxy-3,3',5,5'- tetra-t-butylbiphenyl, a dimer of the 2,4-di-t-butylphenol formed by ortho coupling of
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service