Skip to Content
Merck
All Photos(1)

Key Documents

14462

Sigma-Aldrich

(−)-α-Bisabolol

≥93% (GC)

Synonym(s):

(-)-alpha-Bisabolol, (−)-6-Methyl-2-(4-methyl-3-cyclohexen-1-yl)-5-hepten-2-ol, Levomenol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H26O
CAS Number:
Molecular Weight:
222.37
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥93% (GC)

optical activity

[α]/D -58±5°, neat

refractive index

n20/D 1.496

functional group

hydroxyl

SMILES string

C\C(C)=C\CC[C@](C)(O)[C@H]1CCC(C)=CC1

InChI

1S/C15H26O/c1-12(2)6-5-11-15(4,16)14-9-7-13(3)8-10-14/h6-7,14,16H,5,8-11H2,1-4H3/t14-,15+/m1/s1

InChI key

RGZSQWQPBWRIAQ-CABCVRRESA-N

Looking for similar products? Visit Product Comparison Guide

General description

(-)-α-Bisabolol is the major constituent of essential oil from leaves of Hymenocrater yazdianus.

Application

(-)-α-Bisabolol was used to investigate the leishmanicidal and cytotoxic activity of essential oil of Vanillosmopsis arborea.

Biochem/physiol Actions

α-Bisabolol is active against primary acute leukemia cells, including BCR-ABL(+) acute lymphoblastic leukemias. It is the inhibitor of voltage-dependent Ca(2+) channels in tracheal smooth muscle preparations of rat. It also inhibits human and rat glioma cell growth and survival. It is a potential new therapeutic agent against leishmaniasis.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

289.4 °F - closed cup

Flash Point(C)

143 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Shiva Masoudi et al.
Natural product communications, 7(1), 117-120 (2012-03-21)
Water-distilled essential oils from leaves of Hymenocrater yazdianus Rech.f., flowers of Stachys obtusicrena Boiss., and stems and flowers of Nepeta asterotricha Rech.f., which are endemic to Iran, were analyzed by GC and GC/MS. Fifty-five components of the leaf oil of
Aracélio Viana Colares et al.
Evidence-based complementary and alternative medicine : eCAM, 2013, 727042-727042 (2013-08-13)
The search for new immunopharmacological chemical agents to treat various diseases caused by bacteria, fungi, and protozoa, such as leishmaniasis, for example, has led to the exploration of potential products from plant species and their main active ingredients. Antimonial drugs
Orazio Taglialatela-Scafati et al.
Journal of natural products, 75(3), 453-458 (2012-02-09)
Apart from a large amount (ca. 2.0%) of α-bisabolol β-D-fucopyranoside (2a), the aerial parts of the Mediterranean weed Carthamus glaucus afforded an unusual triglyceride (E-2-crotonyl-1,3-distearolylglycerol, 7), two lipophilic flavonoids (6a,b), and a series of bisabolane fucopyranosides variously acylated on the
Chiaki Nakano et al.
Chembiochem : a European journal of chemical biology, 12(15), 2271-2275 (2012-10-30)
Now found in bacteria: An increasing number of genome sequences indicate that bacteria possess a variety of terpenoid cyclase genes. The characterization of two sesquiterpene cyclase (SC) genes found in the draft genome sequence of Streptomyces citricolor is described here.
Mohamed Attia et al.
Archives of biochemistry and biophysics, 527(1), 37-44 (2012-08-08)
Hernandulcin, a C15 sesquiterpene ketone, is a natural sweetener isolated from the leaves of Lippia dulcis. It is a promising sugar substitute due to its safety and low caloric potential. However, the biosynthesis of hernandulcin in L. dulcis remains unknown.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service