Skip to Content
Merck
All Photos(1)

Key Documents

140961

Sigma-Aldrich

1,2-Dibromopropane

97%

Synonym(s):

Propylene dibromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CHBrCH2Br
CAS Number:
Molecular Weight:
201.89
Beilstein:
1718884
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.519 (lit.)

bp

140-142 °C (lit.)

mp

−55 °C (lit.)

solubility

organic solvents: miscible
water: slightly soluble

density

1.937 g/mL at 25 °C (lit.)

SMILES string

CC(Br)CBr

InChI

1S/C3H6Br2/c1-3(5)2-4/h3H,2H2,1H3

InChI key

XFNJYAKDBJUJAJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Photodissociation dynamics of 1,2-dibromopropane at 234 and 265 nm has been investigated by using velocity map ion imaging method. 1,2-Dibromopropane was used to study the effect of reductant concentration, reductant contact time and suspension pH on reductive dechlorination of carbon tetrachloride by soil manipulated with Fe(II) andHS-. It was used as internal standard during the determination of nitrogenous disinfection byproduct trichloronitromethane by gas chromatography mass spectrometry.

Biochem/physiol Actions

1,2-Dibromopropane induces hepatotoxicity and immunotoxicity in female BALB/c mice.

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

122.0 °F - closed cup

Flash Point(C)

50 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria Jose Cardador et al.
Journal of chromatography. A, 1509, 9-15 (2017-06-19)
This paper described the first analytical method to simultaneously determine 14 disinfection by-products (DBPs) in meat products using microwave-assisted extraction (MAE) and static headspace (SHS) followed by gas chromatography-mass spectrometry (GC-MS). The DBPs included were 4 trihalomethanes, 7 haloacetic acids
S K Lee et al.
Xenobiotica; the fate of foreign compounds in biological systems, 35(1), 97-105 (2005-03-25)
Based on recent results that 1,2-dibromopropane (1,2-DBP) causes hepatotoxicity and immunotoxicity in female BALB/c mice as well as a reduction of hepatic glutathione levels, the possible formation of glutathione conjugates and mercapturic acids of 1,2-DBP was investigated in vivo in
C E Zoetemelk et al.
Drug metabolism and disposition: the biological fate of chemicals, 14(5), 601-607 (1986-09-01)
1,2-Dibromopropane was administered orally in doses of 50-350 mg/kg to male Wistar rats. Four mercapturic acids were identified in urine by GC/MS, viz. N-acetyl-S-(2-oxopropyl)-L-cysteine (I), N-acetyl-S-(2-hydroxypropyl)-L-cysteine (II), N-acetyl-S-(1-carboxyethyl)-L-cysteine (III), and N-acetyl-S-(2-bromo-2-propenyl)-L-cysteine (IV). Mercapturic acid IV was a minor metabolite which
Chun-Sheng Ding et al.
Huan jing ke xue= Huanjing kexue, 34(8), 3113-3118 (2013-11-07)
A novel method is described in this paper, which uses methyl tertiary butyl ether (MTBE) as extractant and 1,2-dibromopropane as internal standard for the determination of nitrogenous disinfection byproduct trichloronitromethane (TCNM) by gas chromatography mass spectrometry (GC-MS). The formation process
Maria Valdivia-Garcia et al.
Scientific reports, 9(1), 9967-9967 (2019-07-12)
Quantitative predictions of impacts on public water supplies are essential for planning climate change adaptations. Monitoring data from five full-scale Scottish drinking water treatment plants (DWTPs) showed that significant correlations exist between conditionally carcinogenic trihalomethanes (THMs) levels, water temperature (r = 0.812

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service