Skip to Content
Merck
All Photos(1)

Documents

138185

Sigma-Aldrich

Hydroxyacetone

contains ≤500 ppm sodium carbonate as stabilizer, technical grade, 90%

Synonym(s):

1-Hydroxy-2-propanone, Acetol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH2OH
CAS Number:
Molecular Weight:
74.08
Beilstein:
605368
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

90%

form

liquid

contains

≤500 ppm sodium carbonate as stabilizer

refractive index

n20/D 1.425 (lit.)

bp

145-146 °C (lit.)

mp

−17 °C (lit.)

solubility

water: miscible

density

1.082 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(=O)CO

InChI

1S/C3H6O2/c1-3(5)2-4/h4H,2H2,1H3

InChI key

XLSMFKSTNGKWQX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hydroxyacetone (Acetol) is important for the manufacture of polyols, acrolein, dyes and skin tanning agents. It undergoes asymmetric reduction to yield (R)-1,2-propanediol in the presence of microbial cell catalyst.

Application

Hydroxyacetone was used to study aerobic oxidation of glycerol and propanediol over metal oxide supported gold nanoparticles in methanol.

Pictograms

FlameHealth hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Muta. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

133.7 °F - closed cup

Flash Point(C)

56.5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hongliang Zhu et al.
Bioresource technology, 149, 238-243 (2013-10-12)
Escherichia coli Lin43 is a strain which has some mutations in glycerol kinase (GlpK) and the repressor for the glycerol 3-phosphate regulon (GlpR). When exposed to glycerol, it quickly accumulates lethal levels of methylglyoxal, which is a precursor of acetol;
I Blank et al.
Advances in experimental medicine and biology, 561, 171-189 (2006-01-28)
The formation of acrylamide (AA) from L-asparagine was studied in Maillard model systems under pyrolysis conditions. While the early Maillard intermediate N-glucosylasparagine generated approximately 2.4 mmol/mol AA, the Amadori compound was a less efficient precursor (0.1 mmol/mol). Reaction with alpha-dicarbonyls
Qi-Xiang Guo et al.
The Journal of organic chemistry, 77(7), 3589-3594 (2012-03-02)
The diastereoselectively switchable enantioselective Mannich reaction of isatin imines with hydroxyacetone is reported. The chiral primary amino acid catalyzed this Mannich reaction to afford both anti- and syn-Mannich adducts in high yields, good diastereoselectivities, and enantioselectivities. The reason for the
Oxidation of glycerol and propanediols in methanol over heterogeneous gold catalysts.
Taarning E, et al.
Green Chemistry, 10(4), 408-414 (2008)
Junsang Ko et al.
Journal of bacteriology, 187(16), 5782-5789 (2005-08-04)
Methylglyoxal (MG) is a toxic metabolite known to accumulate in various cell types. We detected in vivo conversion of MG to acetol in MG-accumulating Escherichia coli cells by use of (1)H nuclear magnetic resonance ((1)H-NMR) spectroscopy. A search for homologs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service