Skip to Content
Merck
All Photos(1)

Documents

136891

Sigma-Aldrich

cis-1,2,3,6-Tetrahydrophthalic anhydride

95%

Synonym(s):

cis-4-Cyclohexene-1,2-dicarboxylic anhydride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8O3
CAS Number:
Molecular Weight:
152.15
Beilstein:
82341
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor density

5.2 (vs air)

Quality Level

vapor pressure

<0.01 mmHg ( 20 °C)

Assay

95%

form

flakes

mp

97-103 °C (lit.)

SMILES string

[H][C@@]12CC=CC[C@]1([H])C(=O)OC2=O

InChI

1S/C8H8O3/c9-7-5-3-1-2-4-6(5)8(10)11-7/h1-2,5-6H,3-4H2/t5-,6+

InChI key

KMOUUZVZFBCRAM-OLQVQODUSA-N

Looking for similar products? Visit Product Comparison Guide

Application

cis-1,2,3,6-Tetrahydrophthalic anhydride (THPA) can be used as:
  • A curing agent for epoxides.
  • A chemical modifier in the modification of polystyrene via Friedel-Crafts acylation for enhancing the thermal stability of the polymer.
  • A reactant for the synthesis of cis-tetrahydroisoindole-1,3-dione derivatives and cyclic diimides.

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

312.8 °F - closed cup

Flash Point(C)

156 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Montmorillonite K-10 supported one-pot synthesis of some symmetric diimides and 3a, 4, 7, 7a-tetrahydroisoindole-1, 3-dione derivatives under solvent-free conditions using microwaves
Habibi D and Marvi O
J. Serb. Chem. Soc., 70(4), 579-583 (2005)
Chemical modification of polystyrene?IV. Electrophilic substitution of polystyrene with cis-1, 2, 3, 6 tetrahydrophthalic anhydride
Biswas M and Chatterjee S
European Polymer Journal, 19(4), 317-320 (1983)
Polymer networks derived from curing of epoxidised linseed oil: influence of different catalysts and anhydride hardeners
Boquillon N and Fringant C
Polymer, 41(24), 8603-8613 (2000)
Influence of cure schedule and stoichiometry on the dynamic mechanical behaviour of tetrafunctional epoxy resins cured with anhydrides
Guerrero P, et al.
Polymer, 37(11), 2195-2200 (1996)
Cong Zhang et al.
Journal of controlled release : official journal of the Controlled Release Society, 226, 193-204 (2016-02-21)
Stepwise pH-responsive nanoparticle system containing charge reversible pullulan-based (CAPL) shell and poly(β-amino ester) (PBAE)/poly(lactic-co-glycolic acid) (PLAG) core is designed to be used as carriers of paclitaxel (PTX) and combretastatin A4 (CA4) for combining antiangiogenesis and chemotherapy to treat hepatocellular carcinoma

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service