Skip to Content
Merck
All Photos(1)

Key Documents

134228

Sigma-Aldrich

5-Methyltryptamine hydrochloride

98%

Synonym(s):

3-(2-Aminoethyl)-5-methylindole hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H14N2 · HCl
CAS Number:
Molecular Weight:
210.70
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

mp

289-292 °C (dec.) (lit.)

solubility

H2O: soluble 50 mg/mL, clear, yellow

functional group

amine

SMILES string

Cl.Cc1ccc2[nH]cc(CCN)c2c1

InChI

1S/C11H14N2.ClH/c1-8-2-3-11-10(6-8)9(4-5-12)7-13-11;/h2-3,6-7,13H,4-5,12H2,1H3;1H

InChI key

RBHDFGBPJGEYCK-UHFFFAOYSA-N

Application

5-Methyltryptamine hydrochloride was used to study the mechanism of metabolism of 9-methyl 1,2,3,4,6,7,12,12b-octahydroindolo[2,3-a]quinolizine in rats. It was used as internal standard in the determination of urinary metabolites of 5-methoxy-N,N-diisopropyltryptamine in humans. It was used as reactant in:
  • synthesis of kinesin spindle protein (KSP) inhibitors
  • intramolecular furan Diels-Alder reactions
  • Pictet-Spengler-like reactions
  • reactant in synthesis of kinesin spindle protein (KSP) inhibitors
  • reactant in intramolecular furan Diels-Alder reactions
  • reactant in Pictet-Spengler-like reactions

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ruan, X., et al.
Huaxue Xuebao, 66, 1731-1731 (2008)
Fokas, D., et al.
Tetrahedron Letters, 44, 5137-5137 (2003)
"Pictet-Spengler-like" Synthesis of Tetrahydro-beta-carbolines under Hydrolytic Conditions. Direct Use of Azalactones as Phenylacetaldehyde Equivalents.
James E. Audia et al.
The Journal of organic chemistry, 61(22), 7937-7939 (1996-11-01)
Tooru Kamata et al.
Drug metabolism and disposition: the biological fate of chemicals, 34(2), 281-287 (2005-11-11)
The urinary metabolites of 5-methoxy-N,N-diisopropyltryptamine (5-MeO-DIPT) in humans have been investigated by analyzing urine specimens from its users. For the unequivocal identification and accurate quantification of its major metabolites, careful analyses were conducted by gas chromatography/mass spectrometry, liquid chromatography/mass spectrometry
A Sparatore et al.
European journal of drug metabolism and pharmacokinetics, 20(2), 135-144 (1995-04-01)
The metabolic fate of 9-methyl 1,2,3,4,6,7,12,12b-octahydroindolo[2,3-a]quinolizine (MIQ), a compound with promising pharmacological action on the CNS system, was investigated in the rat after an oral dose of 200 mg/kg, the maximal tolerated dose. Urine and feces were collected, exhaustively extracted

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service