133337
4-Acetamidobenzoic acid
98%
Synonym(s):
N-Acetyl-PABA
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3CONHC6H4CO2H
CAS Number:
Molecular Weight:
179.17
Beilstein:
390602
EC Number:
MDL number:
UNSPSC Code:
12352106
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
form
powder
reaction suitability
reaction type: solution phase peptide synthesis
mp
259-262 °C (dec.) (lit.)
application(s)
peptide synthesis
SMILES string
CC(=O)Nc1ccc(cc1)C(O)=O
InChI
1S/C9H9NO3/c1-6(11)10-8-4-2-7(3-5-8)9(12)13/h2-5H,1H3,(H,10,11)(H,12,13)
InChI key
QCXJEYYXVJIFCE-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
João Neres et al.
Bioorganic & medicinal chemistry, 15(5), 2106-2119 (2007-01-16)
Benzoic acid and pyridine derivatives inhibit recombinant trans-sialidase from Trypanosoma cruzi with I50 values between 0.4 and 1mM. The best compounds, 4-acetylamino-3-hydroxymethylbenzoic acid and 5-acetylamino-6-aminopyridine-2-carboxylic acid, provide new leads to inhibitors not containing the synthetically complex sialic acid structure. The
Yasuhiro Uno et al.
Biochemical pharmacology, 177, 113996-113996 (2020-04-28)
In humans, polymorphic N-acetyltransferases NAT1 and NAT2 are important enzymes that metabolize endogenous and exogenous compounds, including drugs. These enzymes exhibit considerable inter-individual variability in humans. The cynomolgus macaque is a nonhuman primate species that is widely used in drug
H T Karnes et al.
Journal of chromatography, 338(2), 377-388 (1985-03-22)
A high-performance liquid chromatographic method is described for the analysis of bentiromide metabolites in urine. The procedure involves no more than direct injection of the diluted urine sample, obviating the need for an extraction step or an internal standard. A
M Yasuhara et al.
Biochemical pharmacology, 33(20), 3131-3136 (1984-10-15)
The metabolic and excretory function of the small intestine was investigated after oral and intravenous administration of drugs having an aromatic amino group to rats. After administration of drugs into the intestinal loop at the initial concentration of 0.1 mM
J A Kovacs et al.
Antimicrobial agents and chemotherapy, 37(6), 1227-1231 (1993-06-01)
Isoprinosine has been reported to decrease progression to AIDS, primarily by preventing Pneumocystis carinii pneumonia (PCP), in human immunodeficiency virus-infected patients, but the mechanism of action is unknown. para-Acetamidobenzoic acid (PAcBA), one component of isoprinosine, is structurally related to para-aminobenzoic
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service