131040
Mesaconic acid
99%
Synonym(s):
Methylfumaric acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
HOOCCH=C(CH3)COOH
CAS Number:
Molecular Weight:
130.10
Beilstein:
1722680
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Assay
99%
form
powder
mp
200-202 °C (lit.)
SMILES string
C\C(=C/C(O)=O)C(O)=O
InChI
1S/C5H6O4/c1-3(5(8)9)2-4(6)7/h2H,1H3,(H,6,7)(H,8,9)/b3-2+
InChI key
HNEGQIOMVPPMNR-NSCUHMNNSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
- Process design and economic analysis of renewable isoprene from biomass via mesaconic acid: Details a sustainable approach for producing isoprene, a crucial chemical in the rubber industry, from biomass-derived mesaconic acid, demonstrating its relevance for green chemistry and material science (Lundberg et al., 2019).
- Green Synthesis of a New Al-MOF Based on the Aliphatic Linker Mesaconic Acid: Structure, Properties and In Situ Crystallisation Studies of Al-MIL-68-Mes: Examines the synthesis and properties of a novel metal-organic framework (MOF) utilizing mesaconic acid, applicable in catalysis, gas storage, and separation technologies (Reinsch et al., 2018).
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
R J Truscott et al.
Clinica chimica acta; international journal of clinical chemistry, 110(2-3), 187-203 (1981-03-05)
Two metabolites, 4-hydroxyisovaleric acid and mesaconic acid, have been identified and quantified in the urine of a patient with isovaleric acidemia. These compounds do not appear to have been reported previously as being components of human metabolism. In addition, large
Edward Guccione et al.
Environmental microbiology, 12(3), 576-591 (2009-11-19)
Methylmenaquinol : fumarate reductase (Mfr) is a newly recognized type of fumarate reductase present in some epsilon-proteobacteria, where the active site subunit (MfrA) is localized in the periplasm, but for which a physiological role has not been identified. We show
Yuichiro Himeda et al.
Dalton transactions (Cambridge, England : 2003), (32)(32), 6286-6288 (2009-08-06)
Deuterium-labelled compounds were prepared by (transfer) deuterogenation of unsaturated compounds using H(2) or HCO(2)H in acidic D(2)O as deuterium source with almost quantitative yields and high deuterium contents under mild reaction conditions via heterolytic cleavage of H(2) (or decomposition of
Raúl Alelú-Paz et al.
PloS one, 3(10), e3378-e3378 (2008-10-15)
Immunohistochemical staining of tissues is a powerful tool used to delineate the presence or absence of an antigen. During the last 30 years, antigen visualization in human brain tissue has been significantly limited by the masking effect of fixatives. In
C C Tangney et al.
Neurology, 77(13), 1276-1282 (2011-09-29)
To investigate the interrelations of serum vitamin B12 markers with brain volumes, cerebral infarcts, and performance in different cognitive domains in a biracial population sample cross-sectionally. In 121 community-dwelling participants of the Chicago Health and Aging Project, serum markers of
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service