Skip to Content
Merck
All Photos(1)

Key Documents

128201

Sigma-Aldrich

N-Ethyl-o-crotonotoluidide

97%

Synonym(s):

N-Ethyl-N-(2-methylphenyl)-2-butenamide, Crotamiton

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=CHCON(C2H5)C6H4CH3
CAS Number:
Molecular Weight:
203.28
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

description

predominantly trans

Quality Level

Assay

97%

form

solid

refractive index

n20/D 1.54 (lit.)

bp

153-155 °C/13 mmHg (lit.)

solubility

ethanol: soluble
methanol: soluble

density

0.987 g/mL at 25 °C (lit.)

SMILES string

CCN(C(=O)\C=C\C)c1ccccc1C

InChI

1S/C13H17NO/c1-4-8-13(15)14(5-2)12-10-7-6-9-11(12)3/h4,6-10H,5H2,1-3H3/b8-4+

InChI key

DNTGGZPQPQTDQF-XBXARRHUSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N-Ethyl-o-crotonotoluidide ( Crotamiton) was used to study the photocatalytic decomposition of crotamiton in aqueous solution using TiO2.
N-Ethyl-o-crotonotoluidide (Crotamiton) was used to treat scabies and various pruritic dermatoses.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The use of N-ethyl-o-crotonotoluidide in the treatment of scabies and various pruritic dermatoses.
M COUPERUS
The Journal of investigative dermatology, 13(1), 35-42 (1949-07-01)
Maria Wesołowska et al.
Wiadomosci parazytologiczne, 51(3), 253-256 (2006-08-18)
Demodex mites are common parasites in the hair follicles and in the pilosebaceous glands. Demodex folliculorum and D. brevis have been identified in humans and play a role in three facial conditions: Pityriasis folliculorum, rosacea-like demodicidosis and Demodicidosis gravis. The
E Dika et al.
Cutaneous and ocular toxicology, 25(3), 211-216 (2006-09-19)
Crotamiton is a topical drug used in the treatment of scabies and pruritus. We determined its percutaneous absorption following single and multiple dosing in normal skin. We used in vivo measurement of percutaneous absorption of [14C] crotamiton in a multidose
Noriko Ohtaki et al.
The Journal of dermatology, 30(5), 411-416 (2003-05-30)
We report two cases of scabies treated with oral ivermectin (200 micro g/kg). Case 1, a 72-year-old man, developed crusted scabies with the use of oral corticosteroids due to a misdiagnosis by an earlier physician. The patient was successfully treated
Rika Sekine et al.
Experimental dermatology, 21(3), 201-204 (2012-03-03)
Itch accompanies various skin diseases. As a number of mediators other than histamine can be involved in the itch sensation, H1 receptor antagonists are not necessarily effective in treating itch. External application of antipruritic drugs is occasionally used as an

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service