Skip to Content
Merck
All Photos(2)

Key Documents

127442

Sigma-Aldrich

Aminomethanesulfonic acid

97%

Synonym(s):

(Aminomethyl)sulfonic acid, 1-Aminomethanesulfonic acid, Aminomethanesulfonic acid, Aminomethanesulphonic acid, aminomethylsulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2SO3H
CAS Number:
Molecular Weight:
111.12
Beilstein:
1811756
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

184 °C (dec.) (lit.)

functional group

amine
sulfonic acid

SMILES string

NCS(O)(=O)=O

InChI

1S/CH5NO3S/c2-1-6(3,4)5/h1-2H2,(H,3,4,5)

InChI key

OBESRABRARNZJB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Aminomethanesulfonic acid can be used as a reactant to synthesize: 1-Sulfomethyltetrazole-5-thiol disodium salts, which are useful key intermediates for the preparation of cefonicid sodium. Tridentate N-(2-hydroxybenzyl)aminomethane sulfonic acid Schiff-base ligand by reacting with salicylaldehyde.
It can also be used to functionalize the surface of single-walled carbon nanotubes (SWCNT) or porous metal-organic frameworks (MOF) to introduce the sulfonic acid functional groups for the preparation of nanocomposites.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Copper (II) complexes of Schiff-base and reduced Schiff-base ligands: Influence of weakly coordinating sulfonate groups on the structure and oxidation of 3, 5-DTBC
Sreenivasulu B, et al.
European Journal of Inorganic Chemistry, 2005(22), 4635-4645 (2005)
W S Lewis et al.
The Journal of biological chemistry, 266(31), 20823-20827 (1991-11-05)
The composition and structural aspects of the amino and carboxylic acid groups required for incorporation into peptides by transpeptidation and inhibition of hydrolysis in carboxypeptidase Y-catalyzed reactions were studied. Separation of these two groups by even one carbon prevents incorporation
Sonoko Ishizaki-Koizumi et al.
Biochemical and biophysical research communications, 322(2), 514-519 (2004-08-25)
The activation of Kupffer cells represents a central mechanism of liver injury involving the production of TNF-alpha. It is known that glycine prevents LPS-induced production of TNF-alpha in isolated Kupffer cells. In this study, the possibility that glycine analogues might
Nigel G Ternan et al.
FEMS microbiology letters, 207(1), 49-53 (2002-03-12)
Chromohalobacter marismortui VH1 was screened for its ability to utilise organosulfonate compounds at a range of NaCl concentrations. Only aminomethane sulfonate, of seven sulfonates tested, was utilised. Length of lag phase during growth on aminomethane sulfonate, as either nitrogen and/or
Synthesis of 1-sulfomethyltetrazole-5-thiol disodium salts
Ling Zhang, et al.
Speciality Petrochemicals (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service