11268
Tetraethylammonium bicarbonate
≥95.0% (T)
Synonym(s):
Tetraethylammonium hydrogen carbonate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(CH3CH2)4N(HCO3)
CAS Number:
Molecular Weight:
191.27
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
≥95.0% (T)
form
solid
SMILES string
OC([O-])=O.CC[N+](CC)(CC)CC
InChI
1S/C8H20N.CH2O3/c1-5-9(6-2,7-3)8-4;2-1(3)4/h5-8H2,1-4H3;(H2,2,3,4)/q+1;/p-1
InChI key
XUBMPLUQNSSFHO-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Application
Tetraethylammonium bicarbonate is used:
- In the synthesis of carbamate esters from amines.
- As a precipitant for synthesizing Cu/ZnO catalysts via coprecipitation method.
- As an alternative to the traditional phase-transfer system in [18F]radiofluorinations for preventing microreactor blockages.
- To promote carboxylation of secondary carboxamides bearing a leaving group at the α-position in the synthesis of oxazolidine-2,4-diones.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Monica M Schroll et al.
Journal of the American Society for Mass Spectrometry, 29(10), 2012-2022 (2018-07-19)
Short-term glucose starvation prior to chemotherapy has the potential to preferentially weaken cancer cells, making them more likely to succumb to treatment, while protecting normal cells. In this study, we used 3D cell cultures of colorectal cancer and assessed the
Evaluation of tetraethylammonium bicarbonate as a phase-transfer agent in the formation of [18F] fluoroarenes.
Reed CD, et al.
Journal of Fluorine Chemistry, 143(4), 231-237 (2012)
Use of tetraethylammonium bicarbonate as a precipitation agent on the preparation of coprecipitated Cu/ZnO catalysts.
Jeong C, et al.
Applied Catalysis A: General, 541(4), 35-41 (2017)
Jae-Ung Lee et al.
Physical chemistry chemical physics : PCCP, 22(9), 5057-5069 (2020-02-20)
Graph theory-based reaction pathway searches (ACE-Reaction program) and density functional theory calculations were performed to shed light on the mechanisms for the production of [an + H]+, xn+, yn+, zn+, and [yn + 2H]+ fragments formed in free radical-initiated peptide
Ying Fu et al.
Journal of proteomics, 130, 108-119 (2015-09-19)
Powdery mildew (Pm), caused by Blumeria graminis f. sp. tritici (Bgt), is one of the most important crop diseases, causing severe economic losses to wheat production worldwide. However, there are few reports about the proteomic response to Bgt infection in
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service