Skip to Content
Merck
All Photos(3)

Documents

108898

Sigma-Aldrich

4-Nitro-o-phenylenediamine

98%

Synonym(s):

1,2-Diamino-4-nitrobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H3(NH2)2
CAS Number:
Molecular Weight:
153.14
Beilstein:
608106
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39030501
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

mp

199-201 °C (lit.)

solubility

1 N HCl: ethanol, 1:1: soluble 50 mg/mL

SMILES string

Nc1ccc(cc1N)[N+]([O-])=O

InChI

1S/C6H7N3O2/c7-5-2-1-4(9(10)11)3-6(5)8/h1-3H,7-8H2

InChI key

RAUWPNXIALNKQM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

4-Nitro-o-phenylenediamine was used to study the in vitro activation of 4-nitro-o-phenylenediamine into a potent mutagen in Salmonella typhimurium strain TA98 .

Biochem/physiol Actions

4-Nitro-o-phenylenediamine is a direct-acting mutagen.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marcos Felipe de Oliveira Galvão et al.
Environmental pollution (Barking, Essex : 1987), 233, 960-970 (2017-10-17)
Emissions from burning of biomass in the Amazon region have adverse effects on the environment and human health. Herein, particulate matter (PM) emitted from biomass burning in the Amazon region during two different periods, namely intense and moderate, was investigated.
Y H Shon et al.
Journal of ethnopharmacology, 77(1), 103-109 (2001-08-03)
Extracts from Phellinus linteus, Phellinus igniarius, and Agrocybe cylindracea have been tested for their antimutagenic properties against direct-acting mutagens [4-nitro-o-phenylenediamine (NPD) and sodium azide (NaN(3))] and indirect-acting mutagens [2-aminofluorene (2-AF) and benzo[a]pyrene (B[a]P)], using the Salmonella typhimurium tester strains TA
Talat Baran et al.
International journal of biological macromolecules, 161, 1559-1567 (2020-08-14)
Removal of organic pollutants such as aromatic nitro compounds and dyes from wastewaters is very significant for environment, and the best strategy is their reduction/degradation in the presence of a metal catalyst. In this study, a new palladium nanocatalyst (GA-Sch-Pd)
S J Kaur et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 38(12), 1113-1119 (2000-10-18)
A fraction isolated from Terminalia arjuna was studied for its antimutagenic effect against 4-nitro-o-phenylenediamine (NPD) in TA98, sodium azide in TA100 and 2-aminofluorene (2AF, S9-dependent), a promutagen, in both TA98 and TA 100 tester strains of Salmonella typhimurium using the
J M Gentile et al.
Mutation research, 250(1-2), 79-86 (1991-09-01)
Chlorophyllin, the sodium and copper salt of chlorophyll, chlorophyll a, and chlorophyll b were tested for their ability to inhibit the mutagenic activity of the direct-acting mutagen 4-nitro-o-phenylenediamine (NOP) and its plant-activated mutagenic enhancement. All three forms of chlorophyll were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service