Skip to Content
Merck
All Photos(1)

Key Documents

10879

Sigma-Aldrich

1,1,3-Trichloroacetone

Wacker Chemie AG, produced by Wacker Chemie AG, Burghausen, Germany, ≥86.5% (GC)

Synonym(s):

TCA, 1,1,3-Trichloro-2-propanone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(Cl)2CHCOCH2Cl
CAS Number:
Molecular Weight:
161.41
Beilstein:
1746647
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

produced by Wacker Chemie AG, Burghausen, Germany

Assay

≥86.5% (GC)

manufacturer/tradename

Wacker Chemie AG

bp

88-90 °C/76 mmHg (lit.)

mp

9-11 °C (lit.)

density

1.512 g/mL at 20 °C (lit.)

functional group

chloro
ketone

SMILES string

ClCC(=O)C(Cl)Cl

InChI

1S/C3H3Cl3O/c4-1-2(7)3(5)6/h3H,1H2

InChI key

ZWILTCXCTVMANU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,1,3-Trichloroacetone is direct-acting mutagens in the Ames/Salmonella assay. It is precursor for the generation of 1,3-dihalo oxyallyl intermediates, which undergo [4 + 3] cycloadditions with a host of 1,3-diene systems.

Application

1,1,3-Trichloroacetone was used in base-induced solvolyses of [3.2.1]Bicyclic α,α′-dichloro ketones.

Other Notes

prices for bulk quantities on request

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Base-Induced Solvolyses of [3.2. 1] Bicyclic a, a'-Dichloro Ketones- 1, 3-Transposition and Ring-Contraction.
Fohlisch B, et al.
European Journal of Organic Chemistry, 2001(22), 4357-4365 (2001)
W F Blazak et al.
Mutation research, 206(4), 431-438 (1988-12-01)
1,1,1- and 1,1,3-trichloroacetones (TCA) result from the disinfection of municipal water supplies with chlorine, and are direct-acting mutagens in the Ames/Salmonella assay. The objective of this study was to further investigate the genotoxicity of these compounds in mammalian cells using
Ho Lam Chan et al.
Nature communications, 9(1), 3377-3377 (2018-08-25)
Polycomb repressive complex 1 (PRC1) plays essential roles in cell fate decisions and development. However, its role in cancer is less well understood. Here, we show that RNF2, encoding RING1B, and canonical PRC1 (cPRC1) genes are overexpressed in breast cancer.
M Robinson et al.
Cancer letters, 48(3), 197-203 (1989-12-01)
Several chlorinated acetones have been identified in drinking water and these, as well as a number of chlorinated acroleins, are produced by chlorination of humic acid solutions. Many of these chlorinated compounds and the brominated acrolein analog were positive in
Yusheng Zhang et al.
Science advances, 6(23), eaaz7249-eaaz7249 (2020-06-18)
RING1B, a core Polycomb repressive complex 1 subunit, is a histone H2A ubiquitin ligase essential for development. RING1B is overexpressed in patients with luminal breast cancer (BC) and recruited to actively transcribed genes and enhancers co-occupied by the estrogen receptor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service