Skip to Content
Merck
All Photos(1)

Key Documents

106941

Sigma-Aldrich

2-Methyl-3-nitrophenol

98%

Synonym(s):

2-Hydroxy-6-nitrotoluene, 3-Nitro-o-cresol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(NO2)OH
CAS Number:
Molecular Weight:
153.14
Beilstein:
1946057
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
solid
Assay:
98%

Quality Level

Assay

98%

form

solid

mp

146-148 °C (lit.)

solubility

95% ethanol: soluble 50 mg/mL

functional group

nitro

SMILES string

Cc1c(O)cccc1[N+]([O-])=O

InChI

1S/C7H7NO3/c1-5-6(8(10)11)3-2-4-7(5)9/h2-4,9H,1H3

InChI key

GAKLFAZBKQGUBO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Methyl-3-nitrophenol was used as internal standard to develop a method for measurement of stable isotope ratio of methylnitrophenols in atmospheric particulate matter.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Method for determination of stable carbon isotope ratio of methylnitrophenols in atmospheric particulate matter.
Moukhtar S, et al.
Atmospheric Measurement Techniques, 4(11), 2453-2464 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service