Skip to Content
Merck
All Photos(2)

Key Documents

106437

Sigma-Aldrich

Sodium 1-dodecanesulfonate

ReagentPlus®, ≥99%

Synonym(s):

1-Dodecanesulfonic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11SO3Na
CAS Number:
Molecular Weight:
272.38
Beilstein:
3919536
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Pricing and availability is not currently available. Contact Technical Service

Quality Level

product line

ReagentPlus®

Assay

≥99%

mp

>300 °C (lit.)

solubility

H2O: 25 mg/mL

functional group

sulfonic acid

SMILES string

[Na+]

InChI

1S/C12H26O3S.Na/c1-2-3-4-5-6-7-8-9-10-11-12-16(13,14)15;/h2-12H2,1H3,(H,13,14,15);/q;+1/p-1

InChI key

DAJSVUQLFFJUSX-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium 1-dodecanesulfonate is an ion-pairing reagent and increases the retention of thiabendazole on the HPLC column during determination of thiabendazole in citrus fruit and banana[1]. It is a surfactant standard.

Application

Sodium 1-dodecanesulfonate may be used in synthesis of metal oxide-graphene nanocomposites[2].

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Donghai Wang et al.
ACS nano, 4(3), 1587-1595 (2010-02-27)
Surfactant or polymer directed self-assembly has been widely investigated to prepare nanostructured metal oxides, semiconductors, and polymers, but this approach is mostly limited to two-phase materials, organic/inorganic hybrids, and nanoparticle or polymer-based nanocomposites. Self-assembled nanostructures from more complex, multiscale, and
Y Ito et al.
Journal of chromatography. A, 810(1-2), 81-87 (1998-08-06)
A simple, rapid and reproducible analytical method for thiabendazole (TBZ) and imazalil (IMA) in citrus fruit and banana has been developed. The method involves the use of an ion-exchange cartridge for sample clean-up followed by ion-pair high-performance liquid chromatography with
Chengcheng Yu et al.
Polymers, 11(5) (2019-05-11)
In this work, the effect of doubly functionalized montmorillonite (MMT) on the structure, morphology, thermal, and tribological characteristics of the resulting polystyrene (PS) nanocomposites was investigated. The modification of the MMT was performed using a cationic surfactant and an anionic
D Zielinska et al.
Chemosphere, 40(3), 327-330 (2000-02-09)
The objective of this study was to determine the interaction of modified bilayer lipid membranes (BLMs) with lead and tin organoderivatives. The relative depolarization, of the lipid membrane, caused by organo metals, was used to estimate their activity. Dodecyltrimethylammonium bromide
Manuel Mora et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 78(3), 989-995 (2011-01-14)
An Mg/Al layered double hydroxide (LDH) containing carbonate ion in its interlayer region was examined by medium infrared (MIR) and near infrared reflectance spectroscopy (NIRS). The MIR and NIR spectroscopy techniques was also used to study two organo-hybrid LDHs containing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service