Skip to Content
Merck
All Photos(3)

Documents

104817

Sigma-Aldrich

Trichloro(octadecyl)silane

≥90%

Synonym(s):

Octadecyltrichlorosilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)17SiCl3
CAS Number:
Molecular Weight:
387.93
Beilstein:
1778847
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

vapor density

>1 (vs air)

vapor pressure

<5 mmHg ( 20 °C)

Assay

≥90%

refractive index

n20/D 1.459 (lit.)

bp

223 °C/10 mmHg (lit.)

density

0.984 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCCC[Si](Cl)(Cl)Cl

InChI

1S/C18H37Cl3Si/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-22(19,20)21/h2-18H2,1H3

InChI key

PYJJCSYBSYXGQQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Self-assembled monolayers (SAMs) are used as passivating layers and for controlled modification of surface properties of substrates. Their main applications include wetting, adhesion, friction, chemical sensing and ultra-fine scale lithography. Alkytrichlorosilanes are commonly used for synthesising SAMs on Silicon dioxide which are commonly used in microelectronics. Trichloro(octadecyl)silane is a widely used alkyltrichlorosilane.
Trichloro(octadecyl)silane (OTS) is an alkoxy silane with a large alkyl and polar head group, mainly used as a self-assembled monolayer (SAM) on a variety of nanoparticles and metal oxides substrates. It provides an amphiphilic character that can be used in molecular and opto-electronics applications.

Application

OTS can be used to silanize poly(vinylalcohol) (PVA)/nanosilica (SiO2) nanofibers that can provide a hydrophobic coating on the wooden surfaces by electrospinning. It can also be used as a SAM on the surface of silica (SiO2) gate dielectric for the development of organic field effect transistor (OFETs). It may also be used in a variety of applications like improving the electrochemical performance for dye sensitized solar cells and for the development of metal insulated field effect transistors.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

404.6 °F - closed cup

Flash Point(C)

207 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structural evaluation of 5, 5'-bis (naphth-2-yl)-2, 2'-bithiophene in organic field-effect transistors with n-octadecyltrichlorosilane coated SiO2 gate dielectric.
Ejdrup A, et al.
Langmuir, 34 (23), 6727?6736-6727?6736 (2018)
Surface Characteristics of Silicon Nanowires/Nanowalls Subjected to Octadecyltrichlorosilane Deposition and n-octadecane Coating.
Yilbas BS, et al.
Scientific reports, 6, 38678-38678 (2016)
Fabrication and Characterization of Flexible Metal-Insulator-Semiconductor Field Effect Transistor (MISFET) using Organic ODS and Inorganic ZrO2 as Dielectric Stack Materials.
Haridas PP, et al.
International Journal of Trend in Research and Development, 4, 2394-2394 (2017)
Siyang Yu et al.
Analytical and bioanalytical chemistry, 411(21), 5415-5422 (2019-07-19)
Isoelectric focusing (IEF), a powerful technique for protein separation and enrichment, was successfully integrated into microfluidic paper-based analytical devices (μPADs) in this work. The μPADs for isoelectric focusing were fabricated by octadecyltrichlorosilane (OTS) silanization and subsequent region-selective plasma treatment. The
Shao-Yu Chen et al.
Nano letters, 12(2), 964-969 (2012-01-10)
In this article, we present the transport and magnetotransport of high-quality graphene transistors on conventional SiO(2)/Si substrates by modification with organic molecule octadecyltrichlorosilane (OTS) self-assembled monolayers (SAMs). Graphene devices on OTS SAM-functionalized substrates with high carrier mobility, low intrinsic doping

Articles

Flexible electronic circuits and displays based on organic active materials are future generations of products that may eventually enter mainstream electronics market.

Flexible electronic circuits, displays, and sensors based on organic active materials will enable future generations of electronics products that may eventually enter the mainstream electronics market.

Flexible electronic circuits, displays, and sensors based on organic active materials will enable future generations of electronics products that may eventually enter the mainstream electronics market.

Flexible electronic circuits, displays, and sensors based on organic active materials will enable future generations of electronics products that may eventually enter the mainstream electronics market.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service